Skip to Content
Merck
All Photos(2)

Key Documents

ABE73

Sigma-Aldrich

Anti-FoxP2 Antibody

serum, from rabbit

Synonym(s):

forkhead box P2, trinucleotide repeat containing 10, CAG repeat protein 44, forkhead/winged-helix transcription factor, Trinucleotide repeat-containing gene 10 protein, speech and language disorder 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rabbit, human, mouse

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FOXP2(93986)
mouse ... Foxp2(114142)

General description

Forkhead box protein P2 (FoxP2) is a multidomain transcriptional repressor that belongs to the Fox family of proteins. The FoxP2 protein contains a forkhead-box DNA-binding domain, characteristic of the FOX group of transcription factors. In addition, this protein contains a polyglutamine tract, as well as zinc finger and leucine zipper motifs. In order for FoxP2 to bind DNA, it forms homodimers and heterodimers with FoxP1 and FoxP4.
The forkhead box family of proteins play an important role in immune response, organ development and the development of cancer. Functional studies in vertebrates suggest this with this gene plays a role in the modulation neural plasticity. In line with these findings, mutations of FoxP2 in humans have been linked to speech and language disorders.

Specificity

This antibody recognizes FoxP2.

Immunogen

Epitope: Unknown
GST-tagged recombinant protein corresponding to mouse FoxP2.

Application

Anti-FoxP2 Antibody is a Rabbit Polyclonal Antibody for detection of FoxP2 also known as forkhead box P2, trinucleotide repeat containing 10 & has been validated in WB & IHC.
Research Category
Epigenetics & Nuclear Function

Neuroscience
Research Sub Category
Transcription Factors

Neurodegenerative Diseases
Western Blot Analysis: 1:1,000 dilution from a representative lot detected FoxP2 in 10 µg of L6, NIH/3T3, mouse brain, rat brain, and human brain lysates.

Immunohistochemistry Analysis: A representative lot was used by an independent laboratory in IH. (Lu, M.M., et al. (2002). Mechanisms of Development 119S:S197–S202.)

Quality

Evaluated by Western Blot in RAW264.7 cell lysate.

Western Blot Analysis: 1:1,000 dilution of this antibody detected FoxP2 on 10 µg of RAW264.7 cell lysate.

Target description

80 and 77 kDa Observed. Alternative splicing can result in cell type dependent isoforms.

Physical form

Unpurified
Unpurified rabbit polyclonal serum with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
RAW264.7 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zachary Daniel Burkett et al.
eLife, 7 (2018-01-24)
Human speech is one of the few examples of vocal learning among mammals yet ~half of avian species exhibit this ability. Its neurogenetic basis is largely unknown beyond a shared requirement for FoxP2 in both humans and zebra finches. We
Behavior-linked FoxP2 regulation enables zebra finch vocal learning.
Heston, JB; White, SA
The Journal of Neuroscience null
Zhiliang Fan et al.
Molecular medicine reports, 18(1), 169-178 (2018-05-12)
Epilepsy is a type of refractory neurologic disorder mental disease, which is associated with cognitive impairments and memory dysfunction. However, the potential mechanisms of epilepsy are not well understood. Previous evidence has identified the voltage gated potassium channel complex (VGKC)
Xiaoqin Hu et al.
Experimental and therapeutic medicine, 15(2), 1795-1802 (2018-02-13)
Alzheimer's disease is associated with cognitive impairments that affect memory and executive functions. Simvastatin is a cholesterol-lowering statin drug that is used to control levels of cholesterol in the blood, particularly in cases of hypercholesterolemia, and may be used in
Sam Cooler et al.
Nature neuroscience, 24(1), 105-115 (2020-11-25)
In the vertebrate retina, the location of a neuron's receptive field in visual space closely corresponds to the physical location of synaptic input onto its dendrites, a relationship called the retinotopic map. We report the discovery of a systematic spatial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service