Skip to Content
Merck

ABE490

Anti-NSD2 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Nuclear SET domain-containing protein 2, NSD2, Histone-lysine N-methyltransferase NSD2, Multiple myeloma SET domain-containing protein, MMSET, Protein trithorax-5 and Wolf-Hirschhorn syndrome candidate 1 protein, WHSC1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, IP, WB
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunohistochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... WHSC1(7468)

General description

Nuclear SET domain-containing protein 2 (NSD2) is also called Histone-lysine N-methyltransferase NSD2, Multiple myeloma SET domain-containing protein (MMSET), Protein trithorax-5 and Wolf-Hirschhorn syndrome candidate 1 protein (WHSC1). NSD2 is a histone methyltransferase that acts on histone H3 ′Lys-27′ (H3K27me). Isoform RE-IIBP is thought to be a transcription regulator that uses HADC recruitment to binding DNA and suppress IL5 transcription. Chromosomal aberrations to WHSC1 are linked to multiple myeloma tumors.
~150 kDa observed. Uniprot describes 7 different isoforms ranging from 30 kDa to 152 kDa

Immunogen

KLH-conjugated linear peptide corresponding to a region near the C-terminus of human NSD2.

Application

Detect NSD using this rabbit polyclonal antibody, Anti-NSD2 Antibody validated for use in western blotting, IHC & IP.
Immunohistochemistry Analysis: A 1:500 dilution from a representative lot detected NSD2 in human cerebellum and thalamus tissues.

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected NSD2 in human kidney tissues.

Immunoprecipitation Analysis: 5 µg of this antibody immunoprecipitated NSD2 from 500 µg of HCT-116 cell lysate.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology

Biochem/physiol Actions

This antibody is expected to cross-react with isoforms 1 (152 kDa), 3 (71 kDa), and 4 (66 kDa, REII-BP; IL-5 promoter REII-region-binding protein) based upon immunogen design.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected NSD2 in 10 µg of HeLa cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library