Skip to Content
Merck

Skip To

ABE415

Anti-Transcription factor EB Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Transcripiton factor EB, Class E basic helix-loop-helix protein 35, bHLHe35

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IP, WB
Citations:
-
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

chimpanzee (based on 100% sequence homology), rhesus macaque (based on 100% sequence homology), baboon (based on 100% sequence homology), orangutan (based on 100% sequence homology), gibbon (based on 100% sequence homology)

technique(s)

immunoprecipitation (IP): suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TFEB(7942)

General description

The protein named Transcription factor EB (TFEB) or alternatively in older literature, Class E basic helix-loop-helix protein 35 (bHLHe35) and encoded by the human gene TFEB/BHLH35 is a transcription factor that specifically recognizes and binds E-box sequences (5′-CANNTG-3′) on DNA. Efficient DNA-binding requires dimerization with itself or with another MiT/TFE family member such as TFE3 or MITF. TFEB in association with the protein TFE3, activates the expression of CD40L in T-cells, thereby playing a role in T-cell-dependent antibody responses in activated CD4+ T-cells and thymus-dependent humoral immunity. TFEB also plays a central role in expression of lysosomal genes. TFEB specifically recognizes and binds the CLEAR-box sequence (5′-GTCACGTGAC-3′) present in the regulatory region of many lysosomal genes and through binding acts as a positive regulator of their expression. Finally, TFEB appears to be a positive regulator for autophagy related gene expression, heavy chain immunoglobulin gene expression, and genes necessary for the proper vascularization of important tissues such as the placenta.
~58 kDa and ~40 kDa observed. Uniprot describes two isoforms at 44 kDa and 53 kDa This protein has been observed at 44-53 kDa due to sumoylation (Miller, A. J., et al. (2005). J Biol Chem. 280(1):146-55.).

Immunogen

KLH-conjugated linear peptide corresponding to a region near the C-terminus of human Transcription factor EB.

Application

Detect TFEB using this rabbit polyclonal antibody, Anti-Transcription factor EB Antibody validated for use in western blotting & IP.
Immunoprecipitation Analysis: 10 µg from a representative lot immunoprecipitated Transcription factor EB in 500 µg of Raji cell lysate. Immunopecipitated Transcription factor EB was then detected in Western Blot.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in Raji cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected Transcription factor EB in 10 µg of Raji cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters