Skip to Content
Merck

ABE285

Anti-Tristetraprolin Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Tristetraprolin, TTP, G0/G1 switch regulatory protein 24, Growth factor-inducible nuclear protein NUP475, Protein TIS11A, TIS11, Zinc finger protein 36 homolog, Zfp-36

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ICC, IP, RIP, WB
Citations:
12
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

technique(s)

RIP: suitable, immunocytochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ZFP36(7538)

General description

Tristetraprolin (TTP, Nup 475, TIS11, G0S24, or ZFP36) is a member of the CCCH tandem zinc finger protein family. It is a nuclear RNA-binding protein that functions as a trans-acting factor. It is involved in post-transcriptional regulation of cytokine genes, most notably Tumor Necrosis Factor alpha (TNF-α). Upon activation by lipopolysaccharides, TTP binds TNF-α mRNA at AU-rich domains in the 3′-untranslated Region (3′-UTR) and induces deadenylation, thereby destabilizing the mRNA and tagging it for degradation. To produce this effect, TTP recruits other trans-acting factors to its C-terminal domain and mRNA-decay enzymes to its N-terminal domain. TTP is deactivated by phosphorylation driven by mitogen-activated protein kinase-activated protein kinase 2 (MK2) and p53. Therefore, TTP/MK2 regulates the biosynthesis of TNF-α and mediates inflammatory responses involving TNF-α. Knockout of TTP in mice produces arthritis, alopecia, dermatitis, among a range of other phenotypes related to overexpression of TNF-α.
~40 kDa observed. Uniprot gives a calculated molecular weight of 34 kDa An uncharacterized band appears at ~53 kDa in some lysates.

Immunogen

KLH-conjugated linear peptide corresponding to human Tristetraprolin.

Application

Anti-Tristetraprolin Antibody is an antibody against Tristetraprolin for use in Western Blotting, ICC, Immunoprecipitation, RNA Binding Protein Immunoprecipitation (RIP).
Immunocytochemistry Analysis: A representative lot was used by an independent laboratory to detect Tristetraprolin in human leukocytes. (Brooks, S., et al. (2002). Arthritis and Rheumatism. 46(5):1362–1370.)
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Growth Factors & Receptors

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing PBS without 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
A431 cell lysate
Evaluated by Western Blot in A431 cell lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected Tristetraprolin on 10 µg of A431 cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library