Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IF, IHC, WB
Citations:
-
biological source
rabbit
Quality Segment
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
human
technique(s)
immunofluorescence: suitable, immunohistochemistry: suitable, western blot: suitable
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... TNFRSF17(608)
General description
BCMA (also known as Tumor necrosis factor receptor superfamily member 17, B-cell maturation protein, and CD269) is a receptor of BAFF. BCMA, like TACI, binds BAFF and April. BAFF and its receptors induce B cell development and survival, activate NF-kB and c-Jun N-terminal kinase, and are involved in B cell associated autoimmune diseases.
~27 kDa observed. Uncharacterized bands may be observed in some lysate(s).
Immunogen
Epitope: C-terminus
KLH-conjugated linear peptide corresponding to the C-terminus of human BCMA.
Application
Anti-BCMA Antibody is a highly specific rabbit polyclonal antibody, that targets B-cell Maturation Protein & has been tested in western blotting, IHC & Immunofluorescence.
Immunohistochemistry Analysis: 10 μg/mL from a representative lot detected BCMA in human spleen cells.
Immunofluorescence Analysis: 10 μg/mL from a representative lot detected BCMA in human spleen cells.
Immunofluorescence Analysis: 10 μg/mL from a representative lot detected BCMA in human spleen cells.
Research Category
Apoptosis & Cancer
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
Apoptosis - Additional
Physical form
Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.
Preparation Note
Stable for 1 year at 2-8°C from date of receipt.
Analysis Note
Evaluated by Western Blotting in human spleen, K562, and U937 lysates.
Western Blotting Analysis: 5 μg/mL of this antibody detected BCMA in human spleen, K562, and U937 lysates.
Western Blotting Analysis: 5 μg/mL of this antibody detected BCMA in human spleen, K562, and U937 lysates.
Other Notes
Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| ABC840 | 04053252940644 |