Skip to Content
Merck

ABC67

Anti-Mitochondrial fission 1 protein (Fis1) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

FIS1 homolog, hFis1, Tetratricopeptide repeat protein 11, TPR repeat protein 11

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
3
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FIS1(51024)

General description

Mitochondrial fission 1 protein, hFIs1, is an overexpressed mammlian protein that is localized to the outer mitochondrial membrane of various cell types. It is involved in the fragmentation of the mitochondrial network and its perinuclear clustering, resulting in the release of cytochrome c from the mitochondrion, and apopotosis. It is known to mediate peroxisomal fission and also plays a critical role in histone function by demethylating Lys-4 of histone H3.
~17 kDa observed

Immunogen

Epitope: Cytoplasmic topological domain
KLH-conjugated-linear peptide corresponding to the cytoplasmic topological domain of human Mitochondrial fission 1 protein.

Application

Detect Mitochondrial fission 1 protein (Fis1) using this Anti-Mitochondrial fission 1 protein (Fis1) Antibody validated for use in WB.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Biochem/physiol Actions

Other homologies: Rat and Mouse (83% sequence homology).
This antibody recognizes the cytoplasmic topological domain of Mitochondrial fission 1 protein.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Raji cell lysate
Evaluated by Western Blot in Raji cell lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected Mitochondrial fission 1 protein in 10 µg of Raji cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sofiya Gancheva et al.
Nature communications, 10(1), 4179-4179 (2019-09-15)
The mechanisms underlying improved insulin sensitivity after surgically-induced weight loss are still unclear. We monitored skeletal muscle metabolism in obese individuals before and over 52 weeks after metabolic surgery. Initial weight loss occurs in parallel with a decrease in muscle
Mushfiquddin Khan et al.
Journal of neuroscience research, 96(12), 1900-1913 (2018-07-22)
Traumatic brain injury (TBI) is the major cause of physical disability and emotional vulnerability. Treatment of TBI is lacking due to its multimechanistic etiology, including derailed mitochondrial and cellular energy metabolism. Previous studies from our laboratory show that an endogenous
Zaida Abad-Jiménez et al.
Antioxidants (Basel, Switzerland), 11(7) (2022-07-28)
The chronic low-grade inflammation widely associated with obesity can lead to a prooxidant status that triggers mitochondrial dysfunction. To date, Roux-en-Y gastric bypass (RYGB) is considered the most effective strategy for obese patients. However, little is known about its molecular

Global Trade Item Number

SKUGTIN
ABC6704053252633911

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service