Skip to Content

ABC112

Anti-phospho-ULK1/ATG1 (Ser758) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Serine/threonine-protein kinase ULK1, Autophagy-related protein 1 homolog, ATG1, hATG1, Unc-51-like kinase 1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
polyclonal
Species reactivity:
human
Application:
WB, inhibition assay
Citations:
1
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

bovine (based on 100% sequence homology), rat (based on 100% sequence homology), mouse (based on 100% sequence homology)

technique(s)

inhibition assay: suitable (peptide), western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer758)

Gene Information

human ... ULK1(8408)

General description

ULK1 or autophagy-related protein 1 homolog (ATG1) is a serine/threonine protein kinase. It is a ubiquitously expressed protein that is highly abundant in the brain, skeletal muscle, and heart. It is involved in the signaling pathway of mTOR receptors leading to the regulation of autophagy in nutrient-deprived cells. mTOR may interact with ULK1 as a complex of ULK1/FIP200/Atg13. ULK1 may play an important role in neuronal development: Elimination of ULK1′s enzymatic activity by targeted disruption of its kinase domain has blocked the outgrowth of neurons in the developing mouse brain, possibly by blocking the ULK1-mediated trafficking of neurotrophins.
~120 kDa observed

Immunogen

Epitope: Phosphorylated Ser758
KLH-conjugated linear peptide corresponding to human ULK1/ATG1 phosphorylated at Ser758.

Application

Detect phospho-ULK1/ATG1 (Ser758) using this Anti-phospho-ULK1/ATG1 (Ser758) Antibody validated for use in Western Blotting, Peptide Inhibition Assay.
Peptide Inhibition Analysis: A 1:1,000 dilution from a representative lot blocked phospho-ULK1/ATG1 (Ser758) in serum-starved A431 cell lysates.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Biochem/physiol Actions

This antibody recognizes the ULK1/ATG1 phosphorylated at Ser758.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
A431 and serum starved A431 cell lysates.
Evaluated by Western Blot in A431 and serum starved A431 cell lysates.

Western Blot Analysis: A 1:1,000 dilution of this antibody detected ULK1/ATG1 on 10 µg of A431 and serum starved A431 cell lysates..

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library