Skip to Content
Merck
All Photos(1)

Key Documents

AB9908

Sigma-Aldrich

Anti-QKI-7 Antibody

serum, Chemicon®

Synonym(s):

Quaking Protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, rat, human

manufacturer/tradename

Chemicon®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... QKI(9444)

General description

QKI is an RNA-binding protein that plays a central role in myelinization. Acts by regulating pre-mRNA splicing, mRNA export, mRNA stability and protein translation. It is regulated by alternative splicing.

Specificity

QKI-7 (Quaking Protein). The antibody recognizes the ~42 kDa protein.

Immunogen

Synthetic peptide from mouse QKI-7.

Application

Anti-QKI-7 Antibody is an antibody against QKI-7 for use in IP & WB.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

RNA Binding Protein (RBP)

Physical form

Liquid.
Serum

Storage and Stability

Maintain at -20&deg:C in undiluted aliquots for up to 12 months after date of receipt. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
Positive Control: Glial cells, astrocytes and oligodendrocytes.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiroaki Hojo et al.
The Journal of biological chemistry, 295(2), 390-402 (2019-12-04)
MicroRNA-122 (miR-122) is highly expressed in hepatocytes, where it plays an important role in regulating cholesterol and fatty acid metabolism, and it is also a host factor required for hepatitis C virus replication. miR-122 is selectively stabilized by 3' adenylation
Zhejia Zhang et al.
Oncology letters, 15(4), 5841-5848 (2018-03-21)
Thyroid cancer is the most frequently occurring type of endocrine tumor, with a rapidly increasing incidence rate. MicroRNA (miR)-574-5p is a candidate oncogene in various types of cancer. The present study identified that miR-574-5p affected the cell cycle distribution and
Kristina Sakers et al.
Nature communications, 12(1), 1537-1537 (2021-03-23)
Quaking RNA binding protein (QKI) is essential for oligodendrocyte development as myelination requires myelin basic protein mRNA regulation and localization by the cytoplasmic isoforms (e.g., QKI-6). QKI-6 is also highly expressed in astrocytes, which were recently demonstrated to have regulated
Yutao Wu et al.
Oncotarget, 8(12), 19866-19878 (2017-02-12)
MicroRNA-214(miR-214) has been recently reported to regulate angiogenesis and embryonic stem cells (ESCs) differentiation. However, very little is known about its functional role in vascular smooth muscle cells (VSMCs) differentiation from ESCs. In the present study, we assessed the hypothesis
Xinyun Chen et al.
Nature communications, 12(1), 89-89 (2021-01-06)
The RNA-binding protein QKI belongs to the hnRNP K-homology domain protein family, a well-known regulator of pre-mRNA alternative splicing and is associated with several neurodevelopmental disorders. Qki is found highly expressed in developing and adult hearts. By employing the human

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service