Skip to Content
Merck
All Photos(2)

Key Documents

AB9568

Sigma-Aldrich

Anti-Neurofilament L Antibody

serum, Chemicon®

Synonym(s):

Anti-CMT1F, Anti-CMT2E, Anti-CMTDIG, Anti-NF-L, Anti-NF68, Anti-NFL, Anti-PPP1R110

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, pig, human, bovine, feline, canine, mouse

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... NEFL(4747)

General description

Neurofilaments are a type of intermediate filament that serve as major elements of the cytoskeleton supporting the axon cytoplasm. They are the most abundant fibrillar components of the axon, being on average 3-10 times more frequent than axonal microtubules. Neurofilaments (10 nm in dia.) are built from three intertwined protofibrils which are themselves composed of two tetrameric protofilament complexes of monomeric proteins. The neurofilament triplet proteins (68/70, 160, and 200 kDa) occur in both the central and peripheral nervous system and are usually neuron specific. The 68/70 kDa NF-L protein can self-assemble into a filamentous structure, however the 160 kDa NF-M and 200 kDa NF-H proteins require the presence of the 68/70 kDa NF-L protein to co-assemble. Neuromas, ganglioneuromas, gangliogliomas, ganglioneuroblastomas and neuroblastomas stain positively for neurofilaments. Although typically restricted to neurons, neurofilaments have been detected in paragangliomas and adrenal and extra-adrenal pheochromocytomas. Carcinoids, neuroendocrine carcinomas of the skin, and oat cell carcinomas of the lung also express neurofilaments.

Specificity

Recognizes Neurofilament-Light (NF-L)

Immunogen

Purified porcine NF-L

Application

Detect Neurofilament L using this Anti-Neurofilament L Antibody validated for use in IC, IH, IH(P) & WB.
Research Category
Neuroscience
Research Sub Category
Neurofilament & Neuron Metabolism

Neuronal & Glial Markers
Western Blot Analysis:
1:1000 dilution of a previous lot detected NEUROFILAMENT L on 10 μg of mouse brain lysates.

Immunocytochemistry:
A 1:200-1:500 dilution of a previous lot was used in IC.

Optimal working dilutions must be determined by the end user.

Quality

Immunohistochemistry(paraffin) Analysis:
Neurofilament-L (cat. # AB9568) staining on Normal Cerebellum. Tissue pretreated with Citrate, pH 6.0. This lot of antibody was diluted to 1:800, using IHC-Select Detection with HRP-DAB. Immunoreactivity is seen as fiber-like- staining (brown) around a Purkinje cells.
Optimal Staining With EDTA Buffer, pH 8.0, Epitope Retrieval: Human Cerebellum

Target description

68-70 kDa

Linkage

Replaces: 04-1112

Physical form

Rabbit serum. Contains no preservative.
Unpurified

Storage and Stability

Stable for 1 year at -20ºC from date of receipt.

Analysis Note

Control
Brain tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kerstin Strömvall et al.
PloS one, 12(5), e0176679-e0176679 (2017-05-05)
In order to grow and spread tumors need to interact with adjacent tissues. We therefore hypothesized that small but aggressive prostate cancers influence the rest of the prostate and regional lymph nodes differently than tumors that are more indolent. Poorly
The hematopoietic cytokine granulocyte-macrophage colony stimulating factor is important for cognitive functions.
Krieger, M; Both, M; Kranig, SA; Pitzer, C; Klugmann, M; Vogt, G; Draguhn, A; Schneider, A
Scientific Reports null
Brittney L Gurda et al.
Journal of neuropathology and experimental neurology, 77(3), 229-245 (2018-01-19)
The feline model of Niemann-Pick disease, type C1 (NPC1) recapitulates the clinical, neuropathological, and biochemical abnormalities present in children with NPC1. The hallmarks of disease are the lysosomal storage of unesterified cholesterol and multiple sphingolipids in neurons, and the spatial
NGF activation of TrkA induces vascular endothelial growth factor expression via induction of hypoxia-inducible factor-1?.
Nakamura, K; Tan, F; Li, Z; Thiele, CJ
Molecular and Cellular Neurosciences null
Samantha L Eaton et al.
PloS one, 8(8), e72457-e72457 (2013-09-12)
Western blotting has been a key technique for determining the relative expression of proteins within complex biological samples since the first publications in 1979. Recent developments in sensitive fluorescent labels, with truly quantifiable linear ranges and greater limits of detection

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service