Skip to Content
Merck
All Photos(1)

Key Documents

AB9352

Sigma-Aldrich

Anti-Po Antibody

Chemicon®, from chicken

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

chicken

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Specificity

Recognizes Glycoprotein Po. Po is the major glycoprotein in peripheral myelin

Immunogen

Synthetic peptides from human/mouse Po.

Application

Immunohistochemistry: 1:50-1:100 using 2% paraformaldehyde fixed tissues.

Immunocytochemistry: 1:50-1:100 using 2% paraformaldehyde fixed cells.

Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Neuronal & Glial Markers
This Anti-Po Antibody is validated for use in IC, IH for the detection of Po.

Physical form

Affinity purified chicken IgY. Two different antibodies were combined to make this product. Liquid in PBS, pH 7.2 with 0.02% sodium azide.

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 6 months after date of receipt. It is recommended that the antibody be stored in the dark.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

María A González-González et al.
Scientific reports, 8(1), 16390-16390 (2018-11-08)
Silicone nerve cuff electrodes are commonly implanted on relatively large and accessible somatic nerves as peripheral neural interfaces. While these cuff electrodes are soft (1-50 MPa), their self-closing mechanism requires of thick walls (200-600 µm), which in turn contribute to fibrotic tissue
The 4.1B cytoskeletal protein regulates the domain organization and sheath thickness of myelinated axons.
Einheber, S; Meng, X; Rubin, M; Lam, I; Mohandas, N; An, X; Shrager, P; Kissil, J; Maurel et al.
Glia null
Maria Alejandra Gonzalez-Gonzalez et al.
Frontiers in neuroscience, 16, 726467-726467 (2022-06-03)
Hypertension is a main cause of death in the United States with more than 103 million adults affected. While pharmacological treatments are effective, blood pressure (BP) remains uncontrolled in 50-60% of resistant hypertensive subjects. Using a custom-wired miniature electrode, we
Bradley A Heller et al.
The Journal of cell biology, 204(7), 1219-1236 (2014-04-02)
The PI 3-kinase (PI 3-K) signaling pathway is essential for Schwann cell myelination. Here we have characterized PI 3-K effectors activated during myelination by probing myelinating cultures and developing nerves with an antibody that recognizes phosphorylated substrates for this pathway.
Federica De Angelis et al.
International journal of molecular sciences, 23(23) (2022-12-12)
As a widely prescribed anti-diabetic drug, metformin has been receiving novel attention for its analgesic potential. In the study of the complex etiology of neuropathic pain (NeP), male and female individuals exhibit quite different responses characterized by higher pain sensitivity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service