Skip to Content
Merck
All Photos(1)

Key Documents

AB9080

Sigma-Aldrich

Anti-Ryanodine Receptor 2 Antibody

serum, Chemicon®

Synonym(s):

Anti-ARVC2, Anti-ARVD2, Anti-RYR-2, Anti-RyR, Anti-VACRDS, Anti-VTSIP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human, rat

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... RYR2(6262)

Specificity

Ryanodine Receptor 2.

Immunogen

Synthetic peptide from the variant TM region of human Ryanodine Receptor 2.

Application

Anti-Ryanodine Receptor 2 Antibody is an antibody against Ryanodine Receptor 2 for use in IC, IH & WB.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters

Signaling Neuroscience
Western blot: 1:1,000 using ECL. The antibody reacts with the 500 kDa Ryanodine Receptor 2 protein. Suggested blocking buffer is 10% normal goat serum (or same host as your secondary antibody), 1% BSA in 0.1M PBS with 0.05% Tween 20. Suggested dilution buffer is 1% normal goat serum (or same host as your secondary antibody), 1% BSA in 0.1M PBS with 0.05% Tween 20. Preferred gel percentage is 4-12% gradient gel.

Immunocytochemistry: 1:1,000

Immunohistochemistry: 1:1,000 overnight at 2-8°C using a fluorescently labeled secondary antibody. Suggested fixative is 4% paraformaldehyde in 0.1M PBS (one hour). Suggested permeablization method is 0.05% Triton X-100 in dilution buffer. Suggested blocking buffer is 10% normal goat serum (or same host as your secondary antibody) and 1% BSA in 0.1M PBS. Suggested dilution buffer is 1% normal goat serum (or same host as your secondary antibody) and 1% BSA in 0.1M PBS.

Optimal working dilutions must be determined by the end user.

Physical form

Rabbit serum. Liquid.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
Western blot = adult mouse cardiac muscle or brain (adult mouse skeletal muscle will be negative).

IHC = adult mouse cerebellum Purkinje cells (adult mouse cerebellum granular cells will be negative).

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of CaMKII-mediated phosphorylation of ryanodine receptor type 2 on islet calcium handling, insulin secretion, and glucose tolerance.
Dixit, SS; Wang, T; Manzano, EJ; Yoo, S; Lee, J; Chiang, DY; Ryan, N; Respress et al.
Testing null
Veronica Sacchi et al.
Journal of the American Heart Association, 6(10) (2017-10-12)
Aberrant Ca2+ handling is a prominent feature of heart failure. Elucidation of the molecular mechanisms responsible for aberrant Ca2+ handling is essential for the development of strategies to blunt pathological changes in calcium dynamics. The peptidyl-prolyl cis-trans isomerase peptidyl-prolyl isomerase
Localization and phenotype-specific expression of ryanodine calcium release channels in C57BL6 and DBA/2J mouse strains.
Huang, W; Xing, W; Ryskamp, DA; Punzo, C; Krizaj, D
Experimental Eye Research null
Xiang-Qun Hu et al.
Hypertension (Dallas, Tex. : 1979), 77(4), 1412-1427 (2021-03-02)
[Figure: see text].
Xinyu Yang et al.
Redox biology, 30, 101432-101432 (2020-01-28)
Atrial fibrillation (AF) occurs in up to 11% of cancer patients treated with ibrutinib. The pathophysiology of ibrutinib promoted AF is complicated, as there are multiple interactions involved; the detailed molecular mechanisms underlying this are still unclear. Here, we aimed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service