Skip to Content
Merck
All Photos(1)

Key Documents

AB3560P

Sigma-Aldrich

Anti-Sodium Antibody, Potassium, Chloride Cotransporter 1

Chemicon®, from rabbit

Synonym(s):

Basolateral Na-K-Cl symporter, Bumetanide-sensitive sodium-(potassium)-chloride cotransporter 1, solute carrier family 12 sodium/potassium/chloride transporters), member 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, bovine, mouse, rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SLC12A2(6558)

General description

The Na+/K+/Cl- Cotransporter isoform 1 (NKCC1) is involved in the transport of sodium, potassium, and chloride ions across membranes. NKCC1 mediates sodium and chloride reabsorption and plays an important role in cell volume regulation. NKCC1 is also involved in the control of normal cell proliferation, while its overexpression results in apparent cell transformation, in a manner similar to some protooncogenes (Panet, 1999). NKCC1 is inhibited by bumetanide and is also referred to as the bumetanide-sensitive sodium-(potassium)-chloride cotransporter 1. Inhibition of NKCC1 activity by bumetanide decreases the high-[K+]o-mediated uptake of Cl , blocks astrocyte swelling, and leads to cell shrinkage under isotonic conditions in cells such as retinal pigment epithelia, ventricular myocytes, and vascular endothelial cells (Russell, 2000; Su, 2002). NKCC1 could contribute to an overload of intracellular Na+, K+, and Cl- and cell swelling in conditions such as cerebral ischemia (Yan, 2001). NKCC1 is expressed in multiple tissues with the amount of mRNA expression varying as much as thirty-fold. It is highly expressed in neurons and astrocytes through-out the brain and has been implicated in ischemic necrotic cell death.

Specificity

Expected to cross-react with human and mouse. The immunogen sequence is 95% conserved in mouse, 86% in human, and 82% in bovine.
Recognizes rat Sodium, Potassium, Chloride Cotransporter 1 (NKCC1, BSC2, CCC1). The immunogen shows no significant sequence homology with other CCC.

Immunogen

A 22 amino acid peptide sequence near the C-terminus of rat NKCC1. (Moore-Hoon et al. 1998; Delpire et al. 1994; Payne et al. 1995; Isering et al. 1997; Mount et al. 1998).

Application

Detect Sodium using this Anti-Sodium Antibody, Potassium, Chloride Cotransporter 1 validated for use in ELISA, IH & WB.
Immunohistochemistry:
It is recommended that the antibody be tried at 2-20 µg/mL on paraformaldehyde fixed tissue.

ELISA:
0.5-1.0 µg/mL of a previous lot was used with 1 µg/mL control peptide per well.

Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters

Quality

Evaluated by Western Blot on Rat Spinal Cord lysates.

Western Blot Analysis:
1:500 dilution detected NA+K+CL- COTRANS 1 on 10 µg of Rat Spinal Cord lysates.

Target description

170 kDa

Physical form

Affinity purified rabbit polyclonal in buffer containing PBS with 0.1% BSA.
ImmunoAffinity Purified

Storage and Stability

Stable for 6 months at -20°C in undiluted aliquots from date of receipt.
Handling Recommendations: Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Spinal cord, colon.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mari A Virtanen et al.
Cells, 9(12) (2020-12-10)
Ionotropic GABA transmission is mediated by anion (mainly Cl-)-permeable GABAA receptors (GABAARs). In immature neurons, GABA exerts depolarizing and sometimes functionally excitatory actions, based on active uptake of Cl- by the Na-K-2Cl cotransporter NKCC1. While functional evidence firmly shows NKCC1-mediated
Jichao Chen et al.
Developmental cell, 13(3), 325-337 (2007-09-04)
In the mammalian inner ear, endolymph is produced and resorbed by a complex series of epithelia. We show here that estrogen-related receptor beta (ERR-beta; NR3B2), an orphan nuclear receptor, is specifically expressed in and controls the development of the endolymph-producing
Y Zhu et al.
Neuroscience, 220, 330-340 (2012-06-26)
Persistent inflammation is associated with a shift in spinal GABA(A) signaling from inhibition to excitation such that GABA(A)-receptor activation contributes to inflammatory hyperalgesia. We tested the hypothesis that the primary afferent is the site of the persistent inflammation-induced shift in
Keiichiro Suzuki et al.
Biological & pharmaceutical bulletin, 46(1), 86-94 (2023-01-04)
From our previous observation that the anesthetic effects of phenobarbital potentiate in rats with a decreased cerebral protein expression of the potassium chloride cotransporter KCC2 (SLC12A5), an in vivo study was conducted to clarify whether the pharmacological effect of phenobarbital
Endolymphatic Na? and K? concentrations during cochlear growth and enlargement in mice lacking Slc26a4/pendrin.
Li, X; Zhou, F; Marcus, DC; Wangemann, P
Testing null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service