Skip to Content
Merck
All Photos(1)

Key Documents

AB3081

Sigma-Aldrich

Anti-Na+/H+ Exchanger-1 Antibody

Chemicon®, from rabbit

Synonym(s):

NHE1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, rat, bovine, mouse, pig

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SLC9A1(6548)

Specificity

Recognizes Na+/H+ Exchanger 1 (NHE1). The immunogen peptide shows no significant homology with other NHE isoforms (NHE2-5).

Immunogen

A 22 aa peptide within the cytoplasmic, C-terminal domain of rat NHE1, coupled to KLH.

Application

Detect Na+/H+ Exchanger-1 using this Anti-Na+/H+ Exchanger-1 Antibody validated for use in ELISA & WB.
Western blot: 1-10 μg/mL using Chemiluminescence technique.



ELISA: 1:100,000 using 50-100 ng control peptide per well.

Optimal working dilutions must be determined by end user.

Target description

91 kDa

Physical form

Affinity Purified immunoglobulin. Liquid in PBS and 0.1% BSA .

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hilmi Burak Kandilci et al.
Frontiers in cardiovascular medicine, 7, 617038-617038 (2021-02-16)
Fully-activated Na+/H+ exchanger-1 (NHE1) generates the cardiomyocyte's largest trans-membrane extrusion of H+ ions for an equimolar influx of Na+ ions. This has the desirable effect of clearing excess intracellular acidity, but comes at a large energetic premium because the exchanged
Chao Huang et al.
Journal of neuroscience research, 93(8), 1192-1202 (2015-02-19)
Inducible nitric oxide synthase (iNOS) critically contributes to inflammation and host defense. The inhibition of heat shock protein 70 (Hsp70) prevents iNOS induction in lipopolysaccharide (LPS)-stimulated macrophages. However, the role and mechanism of endogenous Hsp70 in iNOS induction in microglia
Danny Christiansen et al.
Acta physiologica (Oxford, England), 231(3), e13580-e13580 (2020-11-23)
To assess how blood-flow-restricted (BFR) interval-training affects the capacity of the leg muscles for pH regulation during dynamic exercise in physically trained men. Ten men (age: 25 ± 4y; V˙O2max : 50 ± 5 mL∙kg-1 ∙min-1 ) completed a 6-wk interval-cycling intervention (INT) with one
Zhongyan Lu et al.
Journal of dairy science, 99(1), 733-745 (2015-11-09)
Low sodium content in feed and large amounts of salivary sodium secretion are essential requirements to efficient sodium reabsorption in the dairy cow. It is already known that Na(+)/H(+) exchange (NHE) of the ruminal epithelium plays a key role in
Bryan J Feger et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 305(10), R1182-R1189 (2013-09-21)
The myocardial Na(+)/H(+) exchanger-1 (NHE1) plays a major role in regulation of intracellular pH, and its upregulation has been implicated in increased ischemia-reperfusion injury and other pathologies. Hydrogen peroxide (H2O2) increases NHE1 activity acutely via ERK1/2 signaling. Chronic strenuous exercise

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service