Skip to Content
Merck
All Photos(1)

Key Documents

AB1839

Sigma-Aldrich

Anti-Interleukin-6 Antibody

serum, Chemicon®

Synonym(s):

IL-6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

sheep

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

sheep ... Il6(443406)

Specificity

Reacts with ovine IL-6. No reactivity detected to recombinant ovine IL-1beta, IL-8, MCP or TNFalpha.

Immunogen

Recombinant ovine IL-6

Application

Anti-Interleukin-6 Antibody is an antibody against Interleukin-6 for use in ELISA & WB.
Detecting antibody in ELISA, using monoclonal anti-ovine IL-6 as capture antibody. Recommended dilution factor 1:500. See suggested protocol.

Western Blot

Optimal working dilutions must be determined by the end user.

Suggested ELISA Protocol For Ovine IL-6

1. Coat high capacity microtitre wells (e.g. Nunc maxisorp) with monoclonal anti-IL-6 antibody (Chemicon Catalog Number MAB1004) at a concentration of 5 μg/mL in 0.1 M carbonate buffer pH 9.6 overnight at 4ºC.

2. Wash wells 3 x with PBS + 0.05% Tween 20 (PBST).

3. Add samples to wells. Incubate at room temperature in a humidified box for 1 hour. Wash 3 x with PBST.

4. Dilute rabbit anti-IL-6 (Chemicon Catalog Number AB1839) 1:500 in PBST and add 100 μL to wells. Incubate at room temperature for 30 minutes. Wash 3 x with PBST.

5. Dilute anti-rabbit IgG-HRP to appropriate level in PBST and add 100 μL to wells. Incubate 30 minutes at room temperature. Wash 3 x with PBST.

6. Add 100 μL of TMB chromagen to each well. Incubate for 5-10 minutes and stop reaction with 50 μL of 2M sulphuric acid.

Notes:

The anti-rabbit IgG HRP must be adsorbed against mouse immunoglobulin, and should be checked for non-reactivity with coated plates.

Non-specific binding may be reduced by addition of protein such as skim milk powder to the diluents.

Assay sensitivity may be altered by adjusting incubation times and reagent concentrations.
Research Category
Inflammation & Immunology
Research Sub Category
Cytokines & Cytokine Receptors

Linkage

Replaces: 04-1095

Physical form

Rabbit serum. Liquid containing 0.1% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gunlawadee Maneenil et al.
PloS one, 10(3), e0119281-e0119281 (2015-03-21)
A fetal inflammatory response (FIR) in sheep can be induced by intraamniotic or selective exposure of the fetal lung or gut to lipopolysaccharide (LPS). The oral, nasal, and pharyngeal cavities (ONP) contain lymphoid tissue and epithelium that are in contact
Clare A Berry et al.
Reproductive sciences (Thousand Oaks, Calif.), 18(11), 1092-1102 (2011-04-16)
We tested the hypothesis that interleukin 1 (IL-1) mediates intra-amniotic lipopolysaccharide (LPS)-induced chorioamnionitis in preterm fetal sheep. Time-mated Merino ewes with singleton fetuses received IL-1α, LPS, or saline (control) by intra-amniotic injection 1 to 2 days before operative delivery at
Ruth Gussenhoven et al.
Developmental neuroscience, 39(6), 472-486 (2017-08-30)
Chorioamnionitis is associated with adverse neurodevelopmental outcomes in preterm infants. Ureaplasma spp. are the microorganisms most frequently isolated from the amniotic fluid of women diagnosed with chorioamnionitis. However, controversy remains concerning the role of Ureaplasma spp. in the pathogenesis of
Modulation of lipopolysaccharide-induced chorioamnionitis by Ureaplasma parvum in sheep.
Snyder, CC; Wolfe, KB; Gisslen, T; Knox, CL; Kemp, MW; Kramer, BW; Newnham, JP; Jobe et al.
American Journal of Obstetrics and Gynecology null
Charlotte van Gorp et al.
Nutrients, 11(5) (2019-05-01)
Chorioamnionitis, clinically most frequently associated with Ureaplasma, is linked to intestinal inflammation and subsequent gut injury. No treatment is available to prevent chorioamnionitis-driven adverse intestinal outcomes. Evidence is increasing that plant sterols possess immune-modulatory properties. Therefore, we investigated the potential

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service