Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
16
biological source
rabbit
Quality Segment
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
canine, sheep, human
manufacturer/tradename
Chemicon®
technique(s)
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... DDC(1644)
Immunogen
Synthetic peptide from the N-terminal region of human DDC conjugated to KLH.
Application
Anti-Dopa Decarboxylase Antibody is an antibody against Dopa Decarboxylase for use in WB.
Immunoblotting: 1:1000
Optimal working dilutions must be determined by the end user.
Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Neuroscience
Research Sub Category
Neurotransmitters & Receptors
Neuronal & Glial Markers
Neurotransmitters & Receptors
Neuronal & Glial Markers
Biochem/physiol Actions
DOPA Decarboxylase (DDC). Reacts with a single band of ~55 kDa by Western blot. Reacts best with human samples; reactivity low with rodent and rabbit species.
Physical form
Affinity-purified antibody. Liquid in 10 mM HEPES, pH 7.5, 150 mM NaCl, 100 μg/mL BSA and 50% glycerol.
Preparation Note
Maintain at -20 to -80°C in undiluted aliquots for up to six months after date of receipt. Store in a non-self-defrosting freezer. Avoid repeated freeze-thaw cycles.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Explore all of our products under Anti-Dopa Decarboxylase Antibody
— or —
Try our Product Selector Tool to narrow your options
Storage Class
10 - Combustible liquids
wgk
WGK 1
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.