Skip to Content
Merck
All Photos(2)

Key Documents

AB1286-I

Sigma-Aldrich

Anti-Monocarboxylate Transporter 1 Antibody

from chicken, purified by affinity chromatography

Synonym(s):

Protein IGO1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

chicken

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

species reactivity (predicted by homology)

mouse (immunogen homology)

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgY

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

General description

Monocarboxylate Transporter 1 (MCT1), also known as Solute carrier family 16 member 1 (SLC16A1), catalyzes the rapid transport across the plasma membrane of many monocarboxylates such as lactate, pyruvate, branched-chain oxo acids derived from leucine, valine and isoleucine, and the ketone bodies acetoacetate, beta-hydroxybutyrate and acetate. Monocarboxylate Transporter 1 is located in the cell membrane, and is widely expressed in both normal and cancer cells. Monocarboxylate Transporter 1 is involved in symptomatic deficiency in lactate transport (SDLT).

Specificity

This antibody recognizes the C-terminus of Monocarboxylate Transporter 1.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to the C-terminus of rat Monocarboxylate Transporter 1.

Application

Anti-Monocarboxylate Transporter 1 Antibody is a highly specific chicken polyclonal antibody, that targets MCT1 & has been tested in western blotting & IHC (Paraffin).
Immunohistochemistry Analysis: A 1:2,500 dilution from a representative lot detected rat skeletal muscle and rat renal cortex tissues.
Research Category
Neuroscience
Research Sub Category
Signaling Neuroscience

Quality

Evaluated by Western Blot in rat brain membrane tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected rat brain membrane tissue lysate in 10 µg of rat brain membrane tissue lysate.

Target description

Bands are observed at ~44 kDa and ~55 kDa This protein has been observed at ~47 kDa (Sepponen, K., et al. (2003). Acta Physiol Scand. 177(1):79-86.). In addition this protein has been observed at ~55 kDa in certain cell lysates, except in the rumen where two distinct bands appear at ~47 kDa and ~55 kDa (Koho, N., et al. (2005). Comp Biochem Physiol A Mol Integr Physiol. 141(1):29-34.). It is also known to be glycosylated (Carpenter, L., et al. (1996). Biochim Biophys Acta. 1279(2):157-163).

Linkage

Replaces: AB1286

Physical form

Affinity purified
Purified chicken polyclonal IgY in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Rat brain membrane tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gabriel N Valbuena et al.
Molecular neurobiology, 53(4), 2222-2240 (2015-05-13)
Defects in energy metabolism are potential pathogenic mechanisms in amyotrophic lateral sclerosis (ALS), a rapidly fatal disease with no cure. The mechanisms through which this occurs remain elusive and their understanding may prove therapeutically useful. We used metabolomics and stable
Chongliang Zhong et al.
Animal nutrition (Zhongguo xu mu shou yi xue hui), 10, 216-222 (2022-07-06)
Urea nitrogen secreted from blood to rumen is a crucial factor shaping the symbiotic relationship between host ruminants and their microbial populations. Passage of urea across rumen epithelia is facilitated by urea transporter B (UT-B), but the long-term regulation of
Jorge Lopez-Tello et al.
Placenta, 103, 177-179 (2020-11-07)
Twinning and placental fusion occur at a low frequency in species like humans and mice. They are linked to reduced birth weight and intertwin growth discrepancy with important consequences for perinatal survival. Despite this, little is known about the histological
Nigel P Moore et al.
International journal of cell biology, 2016, 2084252-2084252 (2016-11-16)
The distribution of monocarboxylate transporter (MCT) isoforms 1 and 4, which mediate the plasmalemmal transport of l-lactic and pyruvic acids, has been identified in the placentae of rats and rabbits at different ages of gestation. Groups of three pregnant Sprague-Dawley
Chang Cao et al.
Current developments in nutrition, 5(4), nzab025-nzab025 (2021-05-04)
The placenta transports iron to the fetus to support growth and development. Mice are commonly used to study placental iron transport. The subcellular locations of transferrin receptor and ferroportin 1 in iron-transporting cells in the mouse placenta have not been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service