Skip to Content
Merck
All Photos(2)

Key Documents

9601-OP

Sigma-Aldrich

Water

OmniPur® Grade, DEPC Treated, Autoclaved, Nuclease-Free

Synonym(s):

Water

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2O
CAS Number:
Molecular Weight:
18.02
Beilstein:
2050024
MDL number:
UNSPSC Code:
50202301
NACRES:
NA.26
bp:
100 °C (lit.)
Pricing and availability is not currently available.

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

dilution

(for analytical testing)

color

clear colorless

refractive index

n20/D 1.34 (lit.)

bp

100 °C (lit.)

mp

0 °C (lit.)

density

1.000 g/mL at 3.98 °C (lit.)

shipped in

ambient

storage temp.

10-30°C

SMILES string

O

InChI

1S/H2O/h1H2

InChI key

XLYOFNOQVPJJNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Prepared with DEPC for RNA applications. Molecular biology grade.

DNAse: None detected
RNAse: None detected
Protease: None detected





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Warning

Toxicity: Standard Handling (A)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service