Skip to Content
Merck

Skip To

8.55115

Aminomethylated polystyrene LL (100-200 mesh)

Novabiochem®

Synonym(s):

AM resin LL (100-200 mesh)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.22
UNSPSC Code:
13111023
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

Novabiochem®

form

beads

reaction suitability

reaction type: Boc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

amine

storage temp.

2-30°C

General description

Novabiochem® AM resin is prepared according to optimized protocols by direct aminomethylation of polystyrene [1]. This process gives a highly homogeneous and chemically defined support free from potentially reactive chloromethyl groups. This resin can be easily acylated with any appropriate carboxylic acid-containing linker using standard methods of amide bond formation to furnish supports for both Boc and Fmoc batch SPPS and SPOS.

Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins

Analysis Note

Color (visual): white to yellow to beige
Appearance of substance (visual): beads
Loading (determined from the substitution of the Fmoc-Leu loaded resin): 0.30 - 0.50 mmol/g
Swelling Volume (in DMF): lot specific result
TNBS-Test (positive): passes test
TNBS-Test (after loading with Fmoc-Leu-OH (negative)): passes test
The polymer matrix is copoly (styrene - 1% DVB), 100 - 200 mesh.

Other Notes

Replaces: 01-64-0447

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
855020855120855068
description

Novabiochem®

description

Novabiochem®

description

Novabiochem®

description

Novabiochem®

reaction suitability

reaction type: Boc solid-phase peptide synthesis

reaction suitability

reaction type: Boc solid-phase peptide synthesis

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

reaction suitability

reaction type: Boc solid-phase peptide synthesis

functional group

amine

functional group

amine

functional group

amine

functional group

alcohol

product line

Novabiochem®

product line

Novabiochem®

product line

Novabiochem®

product line

Novabiochem®

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

application(s)

peptide synthesis

application(s)

peptide synthesis

application(s)

peptide synthesis

application(s)

peptide synthesis

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters