Skip to Content

8.53036

Boc-Arg(di-Z)-OH

Novabiochem®

Synonym(s):

Boc-Arg(di-Z)-OH, N-α-Boc-N G,N G-bis-CBZ-L-arginine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C27H34N4O8
CAS Number:
Molecular Weight:
542.58
UNSPSC Code:
12352209
NACRES:
NA.22
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

Novabiochem®

assay

≥98% (TLC), ≥98.0% (HPLC)

form

powder

reaction suitability

reaction type: Boc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

mp

140 °C

application(s)

peptide synthesis

storage temp.

15-25°C

SMILES string

N([C@@H](CCCN(\C(=N/C(=O)OCc2ccccc2)\N)C(=O)OCc1ccccc1)C(=O)O)C(=O)OC(C)(C)C

InChI key

FVWNVCDDYWITMG-NRFANRHFSA-N

General description

Building block for introduction of arginine amino-acid residues by Boc SPPS.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Boc SPPS

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Optical rotation α 25/D (c=1 in chloroform): +14.0 - +18.0 °
Purity (TLC(011A)): ≥ 98 %
Purity (TLC(0811)): ≥ 98 %
Assay (HPLC, area%): ≥ 98.0 % (a/a)

To see the solvent systems used for TLC of Novabiochem® products please click here.

Other Notes

Replaces: 04-12-0220

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library