Skip to Content
Merck
All Photos(1)

Documents

8.52089

Sigma-Aldrich

Fmoc-Dpr(Mtt)-OH

Novabiochem®

Synonym(s):

Fmoc-Dpr(Mtt)-OH, N-α-Fmoc-N-β-4-methyltrityl-L-diaminopropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H34N2O4
CAS Number:
Molecular Weight:
582.69
UNSPSC Code:
12352209
NACRES:
NA.22

Quality Level

product line

Novabiochem®

Assay

≥95% (TLC)
≥95.0% (HPLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

amine

storage temp.

15-25°C

InChI

1S/C38H34N2O4/c1-26-20-22-29(23-21-26)38(27-12-4-2-5-13-27,28-14-6-3-7-15-28)39-24-35(36(41)42)40-37(43)44-25-34-32-18-10-8-16-30(32)31-17-9-11-19-33(31)34/h2-23,34-35,39H,24-25H2,1H3,(H,40,43)(H,41,42)/t35-/m0/s1

InChI key

WDZDBCVDBMWMAM-DHUJRADRSA-N

General description

The side-chain Mtt group can be selectively removed with 1% TFA in DCM [1,2,3] or DCM/HFIP/TFE/TES (6.5:2:1:0.5) [4], making this an excellent derivative for the synthesis of branched peptides and peptides modified at the Dpr side-chain [4,5], and for the construction of templates and multifunctionalized resins for combinatorial synthesis[6].

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS

Literature references

[1] K. Barlos, et al., C. H. Schneider & A. N. Eberle (Eds), ESCOM, Leiden, 1993, pp. 283.
[2] A. Aletras, et al. (1995) Int. J. Peptide Protein Res., 45, 488.
[3] L. Bourel, et al. (2000) J. Peptide Sci., 6, 264.
[4] K. Barlos, personal communication.
[5] P. Hoogerhout, et al. (1999) J. Peptide Res., 54, 436.
[6] C. Park & K. Burgess (2001) J. Comb. Chem., 3, 257.

Linkage

Replaces: 04-12-1204

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Optical rotation α 25/D (c=1 in methanol): +10.0 - +13.0 °
Purity (TLC(157A)): ≥ 95 %
Assay (HPLC, area%): ≥ 95.0 % (a/a)
Solubility (1 mmole in 2 ml DMF): clearly soluble

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Novabiochem® offers orthogonally protected amino acids for peptide synthesis, including cyclic and branched peptides.

Novabiochem® offers orthogonally protected amino acids for peptide synthesis, including cyclic and branched peptides.

Novabiochem® offers orthogonally protected amino acids for peptide synthesis, including cyclic and branched peptides.

Novabiochem® offers orthogonally protected amino acids for peptide synthesis, including cyclic and branched peptides.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service