Skip to Content
Merck
All Photos(1)

Key Documents

8.51005

Sigma-Aldrich

DSC

≥99.0% (HPLC), for peptide synthesis, Novabiochem®

Synonym(s):

DSC, N,N′-Disuccinimidyl carbonate, Di-(N-Succinimidyl)carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8N2O7
CAS Number:
Molecular Weight:
256.17
MDL number:
UNSPSC Code:
12352108
EC Index Number:
277-730-3
NACRES:
NA.22
Pricing and availability is not currently available.

Product Name

DSC, N,N′-Disuccinimidyl carbonate Novabiochem®

Quality Level

product line

Novabiochem®

Assay

≥99.0% (HPLC)

form

powder

reaction suitability

reaction type: Coupling Reactions

manufacturer/tradename

Novabiochem®

mp

188-191 °C

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

N2(C(=O)CCC2=O)OC(=O)ON1C(=O)CCC1=O

InChI

1S/C9H8N2O7/c12-5-1-2-6(13)10(5)17-9(16)18-11-7(14)3-4-8(11)15/h1-4H2

InChI key

PFYXSUNOLOJMDX-UHFFFAOYSA-N

General description

A convenient reagent for the synthesis of N-succinimidyl esters of N-protected amino acids [1,2,3,4].

Associated Protocols and Technical Articles
Guide to Selection of Coupling Reagents

Literature references

[1] H. Ogura, et al. (1979) Tetrahedron Lett., 20, 4745.
[2] H. Ogura, et al. (1981) Tetrahedron Lett., 22, 4817.
[3] K. Takeda, et al. (1982) Synth. Commun., 12, 213.
[4] K. Takeda, et al. (1983) TetrahedronLett., 24, 4569.

Application

  • Utilization in solid-phase peptide synthesis: Discusses the role of N,N′-disuccinimidyl carbonate in solid-phase peptide synthesis, providing insights into its effectiveness in enhancing coupling reactions (Meschaninova et al., 2019).
  • Surface functionalization of polymeric materials: This article explores the application of N,N′-disuccinimidyl carbonate in the surface functionalization of polymers to improve their biocompatibility and interaction with biological molecules (Yakufu et al., 2020).
  • Development of affinity chromatography technologies: Details the use of N,N′-disuccinimidyl carbonate in the development of novel affinity chromatography approaches for protein purification (Hinamoto et al., 2023).
  • Enhancement of polymer properties: Investigates how N,N′-disuccinimidyl carbonate can be used to modify polymer surfaces to enhance their properties, such as increased hydrophilicity and reactivity (Swar et al., 2020).

Linkage

Replaces: 01-62-0006

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Assay (HPLC, area%): ≥ 99.0 % (a/a)
Water (K. F.): ≤ 0.50 %
Identity (IR): passes test

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markHealth hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - STOT RE 2 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service