Skip to Content
Merck
All Photos(2)

Key Documents

8.22255

Supelco

Isoamyl alcohol

(mixture of isomers) EMPLURA®

Synonym(s):

Isoamyl alcohol, iso-Pentyl alcohol, 3-Methyl-1-butanol, Iso-Amylalcohol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12O
CAS Number:
Molecular Weight:
88.15
MDL number:
UNSPSC Code:
12352104
EC Index Number:
250-378-8
NACRES:
NB.21
bp:
131 °C/1013 hPa
vapor pressure:
3.1 hPa ( 20 °C)
Pricing and availability is not currently available.

vapor pressure

3.1 hPa ( 20 °C)

Quality Level

product line

EMPLURA®

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

340 °C

expl. lim.

1.2-8 % (v/v)

dilution

(low-cost alternative, for basic applications)

impurities

≤0.3% Water (Karl Fischer)

bp

131 °C/1013 hPa

mp

-117 °C

transition temp

flash point 43 °C

density

0.81 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OCCCCC

InChI

1S/C5H12O/c1-2-3-4-5-6/h6H,2-5H2,1H3

InChI key

AMQJEAYHLZJPGS-UHFFFAOYSA-N

General description

EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA solvents provide adequate specifications with the most important parameters.

Packaging

8.22255.0521: 500ML, Minimum order quantity: 20

Analysis Note

Assay isomers (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.808 - 0.812
Water (K. F.): ≤ 0.30 %
Identity (IR): passes test

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Flam. Liq. 3 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service