8.21029
Phthaloyl dichloride
for synthesis
Synonym(s):
Phthaloyl dichloride, Phthalic acid dichloride
About This Item
Recommended Products
Quality Level
form
liquid
mp
16 °C
transition temp
flash point >110 °C
density
1.41 g/cm3 at 20 °C
storage temp.
2-30°C
SMILES string
ClC(=O)c1c(cccc1)C(=O)Cl
InChI
1S/C8H4Cl2O2/c9-7(11)5-3-1-2-4-6(5)8(10)12/h1-4H
InChI key
FYXKZNLBZKRYSS-UHFFFAOYSA-N
Analysis Note
Density (d 20 °C/ 4 °C): 1.402 - 1.408
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible, corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service