Skip to Content
Merck
All Photos(1)

Key Documents

8.20964

Sigma-Aldrich

Di-n-amyl ether

(stabilised ) for synthesis

Synonym(s):

Di-n-amyl ether, n-Pentyl ether, Amyl ether, Dipentyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2CH2)2O
CAS Number:
Molecular Weight:
158.28
MDL number:
UNSPSC Code:
12352112
EC Index Number:
211-756-8
NACRES:
NA.22
Assay:
≥98.0% (GC)
bp:
186-188 °C/1013 hPa
vapor pressure:
0.7 hPa ( 20 °C)

vapor pressure

0.7 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

180 °C

expl. lim.

2.0-8.0 % (v/v)

bp

186-188 °C/1013 hPa

mp

-69 °C

transition temp

flash point 57 °C

density

0.78 g/cm3 at 20 °C

storage temp.

15-25°C

SMILES string

O(CCCCC)CCCCC

InChI

1S/C10H22O/c1-3-5-7-9-11-10-8-6-4-2/h3-10H2,1-2H3

InChI key

AOPDRZXCEAKHHW-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.783 - 0.788
Peroxide (as H₂O₂): ≤ 0.005 %
Identity (IR): passes test

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

134.6 °F - closed cup

Flash Point(C)

57 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service