Skip to Content
Merck
All Photos(2)

Key Documents

8.20603

Sigma-Aldrich

Glutardialdehyde

(25% solution in water) for synthesis

Synonym(s):

Glutaraldehyde solution, Pentanedial, Glutaric dialdehyde solution, Pentane-1,5-dial

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
OHC(CH2)3CHO
CAS Number:
Molecular Weight:
100.12
Beilstein:
605390
MDL number:
UNSPSC Code:
12352114
NACRES:
NA.22
grade:
for synthesis
synthesis grade
technique(s):
electron microscopy: suitable
bp:
100.5 °C/1013 hPa
vapor pressure:
0.27 hPa ( 20 °C)

grade

for synthesis
synthesis grade

Quality Level

vapor pressure

0.27 hPa ( 20 °C)

form

liquid

concentration

23.0-27.0% (hydrogensulfite method)

technique(s)

electron microscopy: suitable

pH

>3.0 (20 °C in H2O)

bp

100.5 °C/1013 hPa

mp

-6 °C

density

1.06 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[H]C(CCCC([H])=O)=O

InChI

1S/C5H8O2/c6-4-2-1-3-5-7/h4-5H,1-3H2

InChI key

SXRSQZLOMIGNAQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Glutardialdehyde (25% solution in water) can be used in electrochemical studies as a cross-linker chemical to immobilize sensory material on the active region of the electrode and to modify the electrode surface.

Analysis Note

Assay (hydrogensulfit methode): 23.0 - 27.0 %
Density (d 20 °C/ 4 °C): 1.055 - 1.065

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

A novel electrochemical sensor for metoprolol analysis based on glutardialdehyde-zinc oxide modified boron doped diamond electrode
Koccak B, et al.
Diamond and Related Materials, 131, 109558-109558 (2023)
Zhina Hadisi et al.
Macromolecular bioscience, 20(4), e1900328-e1900328 (2020-02-23)
Burn injuries represent a major life-threatening event that impacts the quality of life of patients, and places enormous demands on the global healthcare systems. This study introduces the fabrication and characterization of a novel wound dressing made of core-shell hyaluronic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service