Skip to Content
Merck
All Photos(1)

Documents

8.07489

Sigma-Aldrich

Polyethylene glycol

1500, for synthesis

Synonym(s):

Poly(ethylene glycol), Polyethylene glycol 1500, Polyglycol, Polyethylene oxide, Polyoxy ethylene, PEG 1500, PEG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
13111021
EC Index Number:
500-038-2
NACRES:
NA.22

vapor pressure

<0.01 hPa ( 20 °C)

Quality Level

form

solid

autoignition temp.

420 °C

potency

28000 mg/kg LD50, oral (Rat)
>20000 mg/kg LD50, skin (Rabbit)

pH

4-7 (20 °C, 100 g/L in H2O)

mp

43-49 °C

transition temp

flash point 260 °C

solubility

650 g/L

density

1.2 g/cm3 at 20 °C

bulk density

400‑500 kg/m3

storage temp.

2-30°C

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Polyethylene glycol 1500 can be used as:
  • A plasticizer in the synthesis of polylactic acid/talc composites to improve the crystallinity and mechanical properties of the material.
  • A non-ionic surfactant in the preparation of organoclays.
  • A reactant in the synthesis of quasi-prepolymers by reacting with a polymeric 4,4′-diphenylmethano diisocyanate (pMDI) via the pre-polymerization method.

PEG-1500 can also be used in the aqueous two-phase extraction system along with inorganic salts (sulfates, carbonates, and ammonium phosphates) for the separation and extraction of both organic (amino acids and vitamins) and inorganic substances (metal salts).

Analysis Note

Melting range (lower value): ≥ 43 °C
Melting range (upper value): ≤ 49 °C
Hydroxyl value: 70 - 80
Average molecular mass: 1400 - 1600
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of organoclays using non-ionic surfactants: Effect of time, temperature and concentration
Zampori L, et al.
Applied Clay Science, 48(1-2) (2010)
Extraction of monocarboxylic acids from diluted solutions with polyethylene glycol
Zinov?eva IV, et al.
Theoretical Foundations of Chemical Engineering, 53(5) (2019)
Effect of polyethylene glycol on mechanical, thermal, and morphological properties of talc reinforced polylactic acid composites
Saravana Savitha, et al.
Materials Today: Proceedings, 5(1) (2018)
Analysis and Characterization of Quasi-Prepolymers Obtained from Polyethylene Glycol 1500 and 4, 4-Diphenylmethane-DiIsocyanate
Vazquez NAR, et al.
Material Science and Engineering with Advanced Research, 1(2) (2015)
Recovery of rare-earth elements from nitrate solutions using polyethylene glycol 1500
Fedorova MI, et al.
Russian Chemical Bulletin, 69(7) (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service