8.05817
Magnesium
turnings acc. to Grignard for synthesis
Synonym(s):
Magnesium
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
Mg
CAS Number:
Molecular Weight:
24.31
MDL number:
UNSPSC Code:
12141502
EC Index Number:
231-104-6
NACRES:
NA.22
Quality Level
form
solid
autoignition temp.
>450 °C
bp
1107 °C/1013 hPa
mp
651 °C
density
1.75 g/cm3 at 20 °C
storage temp.
2-30°C
SMILES string
[Mg+2]
InChI
1S/Mg/q+2
InChI key
JLVVSXFLKOJNIY-UHFFFAOYSA-N
Related Categories
Analysis Note
Assay (on Mg): ≥ 99,0 %(m)
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Sol. 1 - Self-heat. 1 - Water-react 2
Storage Class Code
4.2 - Pyrophoric and self-heating hazardous materials
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service