Skip to Content
Merck

8.02407

Quinoline

for synthesis

Synonym(s):

Quinoline, 1-Azanaphthalene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C9H7N
CAS Number:
Molecular Weight:
129.16
UNSPSC Code:
12352005
EC Index Number:
202-051-6
NACRES:
NA.22
MDL number:
Assay:
≥97.0% (GC)
Form:
liquid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

11 Pa ( 25 °C)

Quality Level

assay

≥97.0% (GC)

form

liquid

autoignition temp.

480 °C

potency

270 mg/kg LD50, oral (Rat), 590 mg/kg LD50, skin (Rabbit)

expl. lim.

1.2-7 % (v/v)

pH

7.3 (20 °C, 5 g/L in H2O)

bp

237-238 °C/1013 hPa

mp

-15 °C

transition temp

flash point 107 °C

solubility

6 g/L

density

1.09 g/cm3 at 25 °C

storage temp.

2-30°C

SMILES string

n1c2c(ccc1)cccc2

InChI

1S/C9H7N/c1-2-6-9-8(4-1)5-3-7-10-9/h1-7H

InChI key

SMWDFEZZVXVKRB-UHFFFAOYSA-N

Application


  • Application of quinoline derivatives in third-generation photovoltaics: This article reviews the latest developments in quinoline derivatives, highlighting their implementation in third-generation photovoltaics, showcasing advancements in the field (Lewinska et al., 2021).

  • Recent advances in the synthesis of biologically and pharmaceutically active quinoline and its analogues: A comprehensive review on the synthesis methods of quinoline, discussing their biological and pharmaceutical applications, focusing on innovative synthetic routes (Weyesa et al., 2020).

  • Recent advances in chemistry and therapeutic potential of functionalized quinoline motifs: This review explores the medicinal potential of quinoline and its functionalized derivatives, emphasizing their utility in drug development (Ajani et al., 2022).

  • Comprehensive review on current developments of quinoline-based anticancer agents: This review article covers the role of quinoline compounds in anticancer drug development, highlighting recent progress and new drugs based on the quinoline scaffold (Jain et al., 2019).

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.092 - 1.094
Identity (IR): passes test


Still not finding the right product?

Explore all of our products under Quinoline


signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Chronic 2 - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2

Storage Class

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 2

flash_point_f

224.6 °F

flash_point_c

107 °C



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
802407100004022536373157
802407000504022536373133
802407025004022536373140
802407904904027269240042
802407000004022536373126
802407999904022536373164