Skip to Content
Merck
All Photos(1)

Key Documents

8.01661

Sigma-Aldrich

γ-Butyrolactone

Technipur®, for synthesis

Synonym(s):

γ-Hydroxybutyric acid lactone, 4-Hydroxybutyric acid lactone, GBL

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6O2
CAS Number:
Molecular Weight:
86.09
Beilstein:
105248
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.21
Pricing and availability is not currently available.

grade

for synthesis

Quality Level

vapor density

3 (vs air)

vapor pressure

0.344 hPa ( 20 °C)
1.5 mmHg ( 20 °C)

autoignition temp.

851 °F

expl. lim.

16 %

refractive index

n20/D 1.436 (lit.)

pH

4-5 (20 °C, 100 g/L in H2O)

bp

204-205 °C (lit.)

mp

−45 °C (lit.)

density

1.12 g/mL at 25 °C (lit.)

SMILES string

O=C1CCCO1

InChI

1S/C4H6O2/c5-4-2-1-3-6-4/h1-3H2

InChI key

YEJRWHAVMIAJKC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

γ-Butyrolactone is a common food additive used to enhance the aroma of food.[1] It is used as a solvent for lithium-ion batteries since it has a high flashing point, a low freezing point, a high boiling point, a low viscosity, and a high dielectric constant.[2][3] It is also used as a plasticizer for the solid electrolyte of lithium batteries.[4]

Application

γ-Butyrolactone can be used as:
  • A plasticizer to prepare polymer electrolytes containing poly(epichlorohydrin-ethyleneoxide).[5][6]
  • A plasticizer to prepare quasi-solid-state electrolyte applicable in dye-sensitized solar cells.[7]
  • An electrolyte in combination with ionic liquids for applications in electrochemical devices.[8]

Biochem/physiol Actions

Precursor of γ-hydroxybutyric acid (GHB). It blocks dopamine release by blocking impulse flow in dopaminergic neurons. Pretreatment with γ-butyrolactone allows detection of autoreceptor-induced dopamine release.
Precursor of γ-hydroxybutyric acid (GHB); blocks dopamine release.

Legal Information

Technipur is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel Quasi-Solid-State Electrolyte Based on ?-Butyrolactone and Tetrapropylammonium Iodide for Dye-Sensitized Solar Cells Using Fumed Silica as the Gelling Agent
Senevirathne AMC, et al.
Procedia Engineering, 139, 87-92 (2016)
Phase Equilibrium and Physical Properties for the Purification of Propylene Carbonate (PC) and γ-Butyrolactone (GBL)
Mathuni T, et al.
Journal of Chemical and Engineering Data, 56(1), 89-96 (2011)
Laser Raman and conductivity studies of plasticized polymer electrolyte P(ECH-EO):propylenecarbonate:g-butyrolactone:LiClO4
Nithya H, et al.
Journal of Solid State Chemistry, 16, 1791-1797 (2012)
Ionic liquids and γ-butyrolactone mixtures as electrolytes for supercapacitors operating over extended temperature ranges
Dagousset L, et al.
Royal Society of Chemistry Advances, 5, 13095-13101 (2015)
Effect of propylene carbonate and dimethylformamide on ionic conductivity of P(ECH-EO) based polymer electrolyte
Nithya H, et al.
Electrochimica Acta, 66(1), 110-120 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service