Skip to Content
Merck
All Photos(1)

Key Documents

8.01021

Sigma-Aldrich

3-Bromo-1-propene

(stabilised) for synthesis

Synonym(s):

3-Bromo-1-propene, Allyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2CH=CH2
CAS Number:
Molecular Weight:
120.98
MDL number:
UNSPSC Code:
12352101
EC Index Number:
203-446-6
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

295 °C

potency

120 mg/kg LD50, oral (Rat)

expl. lim.

4.3-7.3 % (v/v)

bp

70-72 °C/1013 mbar

mp

-119 °C

transition temp

flash point -1 °C

density

1.428-1.432 g/cm3 at 20 °C

storage temp.

2-8°C

SMILES string

BrCC=C

InChI

1S/C3H5Br/c1-2-3-4/h2H,1,3H2

InChI key

BHELZAPQIKSEDF-UHFFFAOYSA-N

Application

  • Realizing Both Selective Adsorption and Efficient Regeneration by Adsorbents with Photo-Regulated Molecular Gates: This study employs 3-Bromo-1-propene in the synthesis of a molecular gate mechanism for adsorbents, contributing to selective adsorption and efficient regeneration (J Zhu et al.).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.428 - 1.432
Identity (IR): passes test
The material is stabilised with Butylhydroxytoluene (BHT). At time of analytical testing a content of >100 ppm stabiliser was determined.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

30.2 °F - closed cup

Flash Point(C)

-1 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service