Skip to Content
Merck
All Photos(1)

Key Documents

8.00896

Sigma-Aldrich

Iodoethane

(stabilised with silver) for synthesis

Synonym(s):

Iodoethane, Ethyl iodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H5I
CAS Number:
Molecular Weight:
155.97
MDL number:
UNSPSC Code:
12352101
EC Index Number:
200-833-1
NACRES:
NA.22

vapor pressure

133 hPa ( 18 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

bp

69-73 °C/1013 hPa

mp

-108 °C

transition temp

flash point 53 °C

density

1.93 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

ICC

InChI

1S/C2H5I/c1-2-3/h2H2,1H3

InChI key

HVTICUPFWKNHNG-UHFFFAOYSA-N

Application

  • Photocatalytic degradation of pollutants: Iodoethane is utilized in the synthesis of SnO nanoplates embedded within a chitosan-ionic liquid ionogel framework, enhancing photocatalytic degradation of pollutants through band gap narrowing (Arshia et al., 2023).
  • Amphiphilic ether synthesis: As a reagent in organic synthesis, iodoethane plays a critical role in the production of amphiphilic 6-carboxypullulan ethers, contributing to advancements in carbohydrate polymer applications (Pereira et al., 2014).
  • Detection of pesticide resistance: Iodoethane is employed in assays for detecting glutathione-S transferase activity in mosquitoes, providing a methodological foundation for resistance monitoring in vector control programs (Morou et al., 2010).
  • Synthesis of deuterated compounds: Employed in the synthesis of deuterated sex pheromone components for agricultural pest management, iodoethane facilitates the modification of pheromone structures to improve stability and efficacy (Kiyota et al., 2009).
  • Synthesis of cyanine dyes: Iodoethane is used in the microwave-assisted synthesis of heptamethine cyanine dyes, essential for producing near-infrared dyes with applications in medical imaging and organic electronics (Winstead et al., 2008).

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.910 - 1.940
Ethanol (GC, area%): ≤ 1.0 % (a/a)
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Muta. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

127.4 °F - (External MSDS)

Flash Point(C)

53 °C - (External MSDS)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service