Skip to Content
Merck
All Photos(1)

Key Documents

69041

Sigma-Aldrich

B834(DE3) Competent Cells - Novagen

Escherichia coli, rod shaped

Synonym(s):

Competent cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106202
NACRES:
NA.81

product name

B834(DE3) Competent Cells - Novagen, B834 is the parental strain for BL21. These hosts are methionine auxotrophs and allow high specific activity labeling of target proteins with 35S-methionine and selenomethionine for crystallography.

biological source

Escherichia coli

manufacturer/tradename

Novagen®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

growth mode

adherent or suspension

morphology

rod shaped

technique(s)

microbiological culture: suitable

cell transformation

transformation efficiency: >2×106 cfu/μg

shipped in

dry ice

storage temp.

−70°C

General description

B834 is the parental strain for BL21 (1). These protease-deficient hosts are methionine auxotrophs and allow high specific-activity labeling of target proteins with 35S-methionine and selenomethionine for crystallography (2).

DE3 indicates that the host is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG.
Genotype: F-ompT hsdSB(rB- mB-) gal dcm met (DE3)





This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges us to request certain information about you and the establishment where the GMOs are being handled. Click here for Enduser Declaration (EUD) Form.
B834 is the parental strain for BL21. These hosts are methionine auxotrophs and allow high specific activity labeling of target proteins with 35S-methionine and selenomethionine for crystallography.

Components

0.4 ml1 mlComponent

•2 × 0.2 ml5 × 0.2 mlB834(DE3)Competent Cells

•2 × 2 ml4 × 2 mlSOC Medium

•2 ng2 ngTest Plasmid

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

Other Notes

1. Wood, W. B. (1966) J. Mol. Biol. 16, 118–133.

2. Leahy, D. J., Hendrickson, W. A., Aukhil, I., and Erickson, H. P. (1992) Science 258, 987–991.

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alison B Hickman et al.
eLife, 9 (2020-01-09)
Key to CRISPR-Cas adaptive immunity is maintaining an ongoing record of invading nucleic acids, a process carried out by the Cas1-Cas2 complex that integrates short segments of foreign genetic material (spacers) into the CRISPR locus. It is hypothesized that Cas1
Simon Sander et al.
Protein science : a publication of the Protein Society, 31(6), e4320-e4320 (2022-06-01)
Transient receptor potential melastatin 2 (TRPM2) is a Ca2+ -permeable, nonselective cation channel involved in diverse physiological processes such as immune response, apoptosis, and body temperature sensing. TRPM2 is activated by ADP-ribose (ADPR) and 2'-deoxy-ADPR in a Ca2+ -dependent manner.
Fei Ye et al.
eLife, 7 (2018-12-12)
INAD assembles key enzymes of the Drosophila compound eye photo-transduction pathway into a supramolecular complex, supporting efficient and fast light signaling. However, the molecular mechanism that governs the interaction between INAD and NORPA (phospholipase Cβ, PLCβ), a key step for
Agata Butryn et al.
eLife, 7 (2018-10-06)
Swi2/Snf2 ATPases remodel protein:DNA complexes in all of the fundamental chromosome-associated processes. The single-subunit remodeler Mot1 dissociates TATA box-binding protein (TBP):DNA complexes and provides a simple model for obtaining structural insights into the action of Swi2/Snf2 ATPases. Previously we reported
Anna York et al.
Structure (London, England : 1993), 29(7), 731-742 (2021-03-20)
Branched Lipid II, required for the formation of indirectly crosslinked peptidoglycan, is generated by MurM, a protein essential for high-level penicillin resistance in the human pathogen Streptococcus pneumoniae. We have solved the X-ray crystal structure of Staphylococcus aureus FemX, an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service