Skip to Content
Merck
All Photos(1)

Key Documents

616419-M

Sigma-Aldrich

Transferrin Apo-, Human Plasma, Low Endotoxin Level

Synonym(s):

Transferrin Apo-, Human Plasma, Low Endotoxin Level

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202

Assay

≥95% (SDS-PAGE)

Quality Level

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

Endotoxins, none detected

pI 

5.8

solubility

water: 10 mg/mL

cation traces

Fe: ≤0.01%

storage temp.

−20°C

General description

Native apotransferrin from human plasma. Substantially iron-free. Cell culture tested. Transferrin is a major iron binding protein and is found in human plasma or serum at concentrations of 3 mg/ml. Its concentration correlates with the total iron binding capacity of serum and its level is regulated by the availability of iron. It has bacteriostatic and fungistatic activity and is essential for iron utilization by the hemin-synthesizing reticulocytes. Stored in bone marrow as Tf-bound iron. Has a pI of 5.8.
Native apotransferrin from human plasma. Transferrin is a serum protein involved in iron binding and transport. Substantially iron-free. Cell culture-tested.

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 10 mM phosphate buffer, pH 7.4.

Preparation Note

Prepared from plasma that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 year at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service