Skip to Content
Merck
All Photos(1)

Key Documents

5.89578

Sigma-Aldrich

Toluene

99.8%, anhydrous

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH3
CAS Number:
Molecular Weight:
92.14
Beilstein:
635760
MDL number:
UNSPSC Code:
12191501
NACRES:
NA.22
grade:
anhydrous
Assay:
99.8%
bp:
110-111 °C (lit.)
vapor pressure:
22 mmHg ( 20 °C)
26 mmHg ( 25 °C)
Pricing and availability is not currently available.

grade

anhydrous

Quality Level

vapor density

3.2 (vs air)

vapor pressure

22 mmHg ( 20 °C)
26 mmHg ( 25 °C)

Assay

99.8%

autoignition temp.

997 °F

expl. lim.

7 %

refractive index

n/D 1.496 (lit.)

bp

110-111 °C (lit.)

mp

-93 °C (lit.)

density

0.865 g/mL at 25 °C (lit.)

SMILES string

Cc1ccccc1

InChI

1S/C7H8/c1-7-5-3-2-4-6-7/h2-6H,1H3

InChI key

YXFVVABEGXRONW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.9 °F

Flash Point(C)

4.4 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service