Skip to Content
Merck
All Photos(1)

Key Documents

539741

Sigma-Aldrich

PTP1B Inhibitor

PTP1B Inhibitor, CAS 765317-72-4, is a cell-permeable, selective, reversible and non-competitive allosteric inhibitor of PTP1B (IC₅₀ = 4 µM and 8 µM for PTP1B403 and PTP1B298, respectively).

Synonym(s):

PTP1B Inhibitor, 3-(3,5-Dibromo-4-hydroxy-benzoyl)-2-ethyl-benzofuran-6-sulfonicacid-(4-(thiazol-2-ylsulfamyl)-phenyl)-amide, PTP Inhibitor XXII

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H19Br2N3O7S3
CAS Number:
Molecular Weight:
741.45
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mg/mL
ethanol: 5 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

[S](=O)(=O)(Nc5[s]ccn5)c1ccc(cc1)N[S](=O)(=O)c2cc3[o]c(c(c3cc2)C(=O)c4cc(c(c(c4)Br)O)Br)CC

InChI

1S/C26H19Br2N3O7S3/c1-2-21-23(24(32)14-11-19(27)25(33)20(28)12-14)18-8-7-17(13-22(18)38-21)41(36,37)30-15-3-5-16(6-4-15)40(34,35)31-26-29-9-10-39-26/h3-13,30,33H,2H2,1H3,(H,29,31)

InChI key

SXKBTDJJEQQEGE-UHFFFAOYSA-N

General description

A cell-permeable, selective, reversible, and non-competitive allosteric inhibitor of PTP1B (IC50 = 4 µM and 8 µM for PTP1B403 and PTP1B298, respectively). Binds to a novel site away from the catalytic pocket and inhibits PTP1B activity by preventing closure of the WPD loop. Also available in InSolution format (Cat. No. 539749.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PTP1B403
Product does not compete with ATP.
Reversible: yes
Target IC50: 4 µM and 8 µM for PTP1B403 and PTP1B298, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Wiesmann, C., et al. 2004. Nat. Struct. Mol. Biol.11, 730.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vitor Ferreira et al.
Redox biology, 63, 102741-102741 (2023-05-26)
Olanzapine (OLA), a widely used second-generation antipsychotic (SGA), causes weight gain and metabolic alterations when administered orally to patients. Recently, we demonstrated that, contrarily to the oral treatment which induces weight gain, OLA administered via intraperitoneal (i.p.) in male mice
Hyo Jeong Kim et al.
Frontiers in neuroscience, 15, 725398-725398 (2021-10-26)
Rett syndrome (RTT) is a severe X-linked dominant neurodevelopmental disorder caused by mutations in the methyl-CpG-binding protein 2 (MECP2) gene; MeCP2 regulates the expression of brain-derived neurotrophic factor (BDNF) and increasing BDNF levels ameliorates RTT symptoms. However, the clinical application
Bo Gong et al.
Nature, 631(8020), 415-423 (2024-06-13)
Directed cell migration is driven by the front-back polarization of intracellular signalling1-3. Receptor tyrosine kinases and other inputs activate local signals that trigger membrane protrusions at the front2,4-6. Equally important is a long-range inhibitory mechanism that suppresses signalling at the
Sarah Zierold et al.
Journal of the American Heart Association, 10(6), e018322-e018322 (2021-03-06)
Background Brain-derived neurotrophic factor (BDNF) is expressed in neuronal and nonneuronal cells and may affect vascular functions via its receptor, tropomyosin-related kinase B (TrkB). In this study, we determined the expression of BDNF in different perivascular adipose tissue (PVAT) depots

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service