Skip to Content
Merck
All Photos(1)

Key Documents

5.30389

Sigma-Aldrich

GSK′872

≥98% (HPLC), solid, RIPK3 inhibitor, Calbiochem®

Synonym(s):

RIPK3 Inhibitor, GSK′872, N-(6-(Isopropylsulfonyl)quinolin-4-yl)benzo[d]thiazol-5-amine, RIP3 Kinase Inhibitor I, Receptor-Interacting Protein Kinase 3 Inhibitor I, GSK-872, Necrosis Inhibitor V, RIPK3 Inhibitor I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H17N3O2S2
CAS Number:
Molecular Weight:
383.49
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

RIPK3 Inhibitor, GSK′872,

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(C(C)C)c1cc2c([nH]cc[c]2=Nc3cc4nc[s]c4cc3)cc1

InChI

1S/C19H17N3O2S2/c1-12(2)26(23,24)14-4-5-16-15(10-14)17(7-8-20-16)22-13-3-6-19-18(9-13)21-11-25-19/h3-12H,1-2H3,(H,20,22)

InChI key

ZCDBTQNFAPKACC-UHFFFAOYSA-N

General description

A cell-permeable quinolinyl-benzothiazolamine compound that is reported to act as a RIP3-selective kinase inhibitor with >1,000-fold selectivity over a vast majority of more than 300 other kinases, including RIP1. RIP1 inhibitor Nec-1 (30 µM; Cat. Nos. 480065 & 505224 ) and genetic knockdown can be employed in conjunction with GSK′872 (3 µM) in delineating RIP1- vs. RIP3-dependency in Toll-like receptors- (TLRs) mediated necrosis and TNFR1-initiated necroptosis in the presence or absence of caspase inhibitor Z-VAD-FMK (25 µM; Cat. Nos. 627609 & 627610 ). While RIP3 and its downstream substrate MLKL are shown to mediate TLR3-initiated necrosis in the presence of Z-VAD-FMK in murine macrophage (BMDM & J774), fibroblast (3T3-SA & MEF), and endothelial (SVEC4-10) cultures, only in BMDM & J774 macrophage cultures is RIP1 involvement also seen in TLR3-mediated necrosis.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
RIP3 Kinase
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze 9-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Kaiser, W.J., et al. 2013. J. Biol. Chem.288, 31268.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lei Wu et al.
Cancer immunology research, 8(5), 710-721 (2020-03-04)
Metabolic reprogramming is critical for the polarization and function of tumor-associated macrophages (TAM) and hepatocarcinogenesis, but how this reprogramming occurs is unknown. Here, we showed that receptor-interacting protein kinase 3 (RIPK3), a central factor in necroptosis, is downregulated in hepatocellular
Yun Wang et al.
Frontiers in cellular and infection microbiology, 9, 286-286 (2019-08-24)
Programmed cell death and especially necroptosis, a programmed and regulated form of necrosis, have been recently implicated in the progression and outcomes of influenza in mouse models. Moreover, Z-DNA/RNA binding protein 1 (ZBP1) has been identified as a key signaling

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service