Skip to Content
Merck
All Photos(1)

Key Documents

516360

Sigma-Aldrich

Pepsin, Porcine Stomach Mucosa

Catalyzes the hydrolysis of aminoacyl-proline to an amino acid and proline. Inhibitors include aliphatic alcohols, pepstatin A, and pH 6.0.

Synonym(s):

Pepsin, Porcine Stomach Mucosa

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.25
Pricing and availability is not currently available.

Quality Level

form

lyophilized

specific activity

≥2000 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

solubility

10 mM HCl: 0.5 mg/mL

shipped in

ambient

storage temp.

−20°C

Related Categories

General description

Native pepsin from porcine stomach mucosa. Active under acidic conditions. Catalyzes the hydrolysis of aminoacyl-proline to an amino acid and proline. Inhibitors include aliphatic alcohols, pepstatin A, and pH >6.0. Has an optimal pH of 2.0-4.0.
Native pepsin from porcine stomach mucosa. Secreted by the mucosal lining of the stomach. Active under acidic conditions. Catalyzes the hydrolysis of aminoacyl-proline to an amino acid and proline. Inhibitors include aliphatic alcohols, pepstatin A, and pH >6.0.

Warning

Toxicity: Harmful (C)

Unit Definition

One unit is defined as the amount of enzyme that will digest hemoglobin substrates to TCA-soluble products, measured as a change in absorbance at 280 nm of 0.001 per min at 37°C, pH 2.0.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Bergmeyer, H.V. 1984. Methods of Enzymatic Analysis (3rd ed., vol. 5) p. 232, Academic Press, New York.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service