Skip to Content
Merck
All Photos(1)

Key Documents

5.08741

Millipore

DAPI Stain

Synonym(s):

DAPI Stain, 4ʹ,6-Diamidino-2-phenylindole, 2HCl, 2-(4-Amidinophenyl)-1H-indole-6-carboxamidine, 2HCl, 2-(4-Carbamimidoylphenyl)-1H-indole-6-carboximidamide, 2HCl, DAPI, ≥97% by HPLC, in H₂O

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H15N5 · 2HCl
CAS Number:
Molecular Weight:
350.25
MDL number:
UNSPSC Code:
41116130
NACRES:
NA.21
Pricing and availability is not currently available.

Assay

≥97% (HPLC)

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
protect from light

fluorescence

λex ~359 nm
λem ~461 nm

storage temp.

−20°C

SMILES string

Cl.Cl.[nH]1c2c(cc1c3ccc(cc3)\C(=N\[H])\N)ccc(c2)\C(=N\[H])\N

InChI

1S/C16H15N5.2ClH/c17-15(18)10-3-1-9(2-4-10)13-7-11-5-6-12(16(19)20)8-14(11)21-13;;/h1-8,21H,(H3,17,18)(H3,19,20);2*1H

InChI key

FPNZBYLXNYPRLR-UHFFFAOYSA-N

General description

Cell-permeable DNA-binding dye. Binds preferentially to DNA rich in adenine and thymine. Used in flow cytometry for measuring nuclear DNA content or for sorting isolated chromosomes. Useful for microscopic detection of nuclei and nuclear DNA in normal and apoptotic cells. Can also be used to detect mycoplasma. A reversible inhibitor of S-adenosyl-L-methionine decarboxylase and diamine oxidase. Cells are typically stained at 1 µg/ml.
Cells are typically stained at 1 µg/ml.

Application


  • Determination of Ploidy Levels and Nuclear DNA Content in Cryptococcus neoformans by Flow Cytometry: Drawbacks with Variability: This research utilizes DAPI for quantifying nuclear DNA content and determining ploidy levels in Cryptococcus neoformans, providing critical insights into fungal genetics and potential challenges in reproducibility (Chang et al., 2024).

  • Near-Infrared Light-Excited Quinolinium-Carbazole Small Molecule as Two-Photon Fluorescence Nucleic Acid Probe: Highlights the development of a novel two-photon fluorescence probe for nucleic acid imaging, employing DAPI as a reference for comparing fluorescence characteristics, enhancing molecular imaging techniques (Sun et al., 2024).

  • A FACS-based Novel Isolation Technique Identifies Heterogeneous CTCs in Oral Squamous Cell Carcinoma: Applies DAPI for the enumeration and characterization of circulating tumor cells (CTCs), showcasing its importance in cancer diagnostics and the potential for personalized medicine approaches (Chauhan et al., 2024).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
DNA
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Physical form

A 5 mg/ml (14.27 mM) sterile-filtered solution of DAPI, 2HCl (Cat. No. 268298) in H₂O.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C). Aliquots are stable for up to 6 months at -20°C.

Other Notes

Mathieu, C., et al. 1995. Exp. Cell Res.216, 13.
Cubria, C., et al. 1994. Comp. Biochem. Physiol. C.105, 251.
Hotz, M.A., et al. 1994. Cytometry15, 237.
Lee, L.S., and Garnett, H.M. 1993. J. Biochem. Biophys. Methods26, 249.
Miller, T., et al. 1993. Biotechniques15, 1042.

Gray, J.W., et al. 1982. Science238, 323.
Hessling, J.J., et al. 1980. J. Immunol. Methods38, 315.

Russels, W.C., et al. 1975. Nature253, 461.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service