Skip to Content
Merck

5.00090

L(+)Ascorbyl palmitate

EMPROVE® ESSENTIAL, Ph. Eur., BP, NF, FCC, E 304

Synonym(s):

L(+)Ascorbyl palmitate, Palmitoyl-L-ascorbic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C22H38O7
CAS Number:
Molecular Weight:
414.53
UNSPSC Code:
12352108
E Number:
E304
EC Index Number:
205-305-4
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

agency

BP, NF, Ph. Eur.

reg. compliance

FCC

product line

EMPROVE® ESSENTIAL

form

solid

potency

>5000 mg/kg LD50, oral (Rat)

mp

107-117 °C

application(s)

pharmaceutical
semi-solid formulation
solid formulation

storage temp.

15-25°C

InChI

1S/C22H38O7/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-18(24)28-16-17(23)21-19(25)20(26)22(27)29-21/h17,21,23,26-27H,2-16H2,1H3

InChI key

NULLRXHRYOXSEW-UHFFFAOYSA-N

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Ascorbyl palmitate is typically used as an antioxidant in food and pharmaceutical applications.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
500090250004022536255828
500090901004022536681139