499643
Mucin, Bovine Submaxillary Gland
Native mucin from bovine submaxillary gland. It is a rich source of O-linked oligosaccharide chains and it is suitable as a substrate for neuraminidase.
Synonym(s):
Mucin, Bovine Submaxillary Gland
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
description
Merck USA index - 14, 6296
Quality Level
form
lyophilized
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
desiccated
solubility
dilute aqueous base buffer: soluble
water: soluble
shipped in
ambient
storage temp.
2-8°C
Related Categories
General description
Native mucin from bovine submaxillary gland. A high molecular weight glycoprotein that is the major constituent of saliva, gastrointestinal juices and other secretions. Mucin exists as an aggregated complex in excess of 106 molecular weight. It is a rich source of O-linked oligosaccharide chains and it is suitable as a substrate for neuraminidase.
Native mucin from bovine submaxillary gland. A high molecular weight glycoprotein that is the major constituent of saliva, gastrointestinal juices, and other secretions. Mucin exists as an aggregated complex in excess of 106 molecular weight. It is a rich source of O-linked oligosaccharide chains and it is suitable as a substrate for neuraminidase.
Warning
Toxicity: Standard Handling (A)
Reconstitution
Following reconstitution, refrigerate (4°C).
Other Notes
Straus, D.C., et al. 1996. Infect. Immun. 64, 1446.
Savage, A.V., et al. 1990. Eur. J. Biochem. 192, 427.
Savage, A.V., et al. 1990. Eur. J. Biochem. 192, 427.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service