Skip to Content
Merck
All Photos(1)

Key Documents

4610-OP

Millipore

Formamide

OmniPur® Grade, >= 99%, Deionized

Synonym(s):

Formamide, Carbamaldehyde, Amide C1, Formic amide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCONH2
CAS Number:
Molecular Weight:
45.04
Beilstein:
505995
MDL number:
UNSPSC Code:
12352111
NACRES:
NA.21
technique(s):
hybridization: suitable
bp:
210 °C (lit.)
vapor pressure:
0.08 mmHg ( 20 °C)
30 mmHg ( 129 °C)
Pricing and availability is not currently available.

vapor density

1.55 (vs air)

Quality Level

vapor pressure

0.08 mmHg ( 20 °C)
30 mmHg ( 129 °C)

Assay

≥99% (GC)

form

viscous liquid (slightly)

autoignition temp.

932 °F

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

technique(s)

hybridization: suitable

color

colorless

refractive index

n20/D 1.447 (lit.)

pH

8-10 (20 °C, 200 g/L)

bp

210 °C (lit.)

mp

2-3 °C (lit.)

density

1.134 g/mL at 25 °C (lit.)

shipped in

wet ice

storage temp.

2-8°C

SMILES string

NC=O

InChI

1S/CH3NO/c2-1-3/h1H,(H2,2,3)

InChI key

ZHNUHDYFZUAESO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description



Molecular Biology grade.

A260 (0.5 M, water): ≤0.03 AU
A270 (0.5M, water): ≤0.05 AU
A280 (0.5 M, water): ≤0.08 AU
Purity by Assay: ≥ 99%
Conductivity: ≤100 µmho
DNase: None detected
Heavy metals (as Pb): ≤0.0001%
Identification: To pass test
Phosphatase: None detected
Protease: None detected
RNase: None detected





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Application

Formamide destabilizes nucleic acid duplexes and may be used, typically, at a concentration of 50%, in hybridization protocols requiring lower hybridization temperatures.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 1B - STOT RE 2 Oral

Target Organs

Blood

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

305.6 °F

Flash Point(C)

152 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service