Skip to Content
Merck
All Photos(1)

Key Documents

422500

Sigma-Aldrich

Kifunensine, Kitasatosporia kifunense

A potent alkaloid inhibitor of mannosidase I.

Synonym(s):

Kifunensine, Kitasatosporia kifunense

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12N2O6
CAS Number:
Molecular Weight:
232.19
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

hot water: 25 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

N21[C@H](NC(=O)C2=O)[C@H]([C@H]([C@@H]([C@H]1CO)O)O)O

InChI

1S/C8H12N2O6/c11-1-2-3(12)4(13)5(14)6-9-7(15)8(16)10(2)6/h2-6,11-14H,1H2,(H,9,15)/t2-,3-,4+,5+,6+/m1/s1

InChI key

OIURYJWYVIAOCW-PQMKYFCFSA-N

General description

A potent alkaloid inhibitor of mannosidase I. An ineffective inhibitor of mannosidase II and the endoplasmic reticulum α-mannosidase.
A potent alkaloid inhibitor of mannosidase I. Does not affect mannosidase II and the endoplasmic reticulum α-mannosidase.

Biochem/physiol Actions

Cell permeable: no
Primary Target
mannosidase-1
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Weng, S., and Spiro, R.G. 1996. Arch. Biochem. Biophys.325, 113.
Kayakiri, H., et al. 1991. Chem. Pharm. Bull. 39, 1378.
Elbein, A.D., et al. 1990. J. Biol. Chem. 265, 15599.
Iwami, M., et al. 1987. J. Antibiotic. 40, 612.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Merve Sen et al.
Biomolecules, 11(10) (2021-10-24)
Rhodopsin (RHO) misfolding mutations are a common cause of the blinding disease autosomal dominant retinitis pigmentosa (adRP). The most prevalent mutation, RHOP23H, results in its misfolding and retention in the endoplasmic reticulum (ER). Under homeostatic conditions, misfolded proteins are selectively

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service