Skip to Content
Merck

Skip To

4160

L-Isoleucine

An essential branched chain amino acid.

Synonym(s):

L-Isoleucine, Ile

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C6H13NO2
CAS Number:
Molecular Weight:
131.17
UNSPSC Code:
12352209
NACRES:
NA.25
MDL number:
eCl@ss:
32160406
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

description

RTECS - NR4705000
Merck USA index - 14, 5179

assay

≥99% (TLC)

form

crystalline solid, crystals

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: ~4% (at RT)

shipped in

ambient

storage temp.

15-25°C

SMILES string

[N+H3][C@@H]([C@H](CC)C)C(=O)[O-]

InChI

1S/C6H13NO2/c1-3-4(2)5(7)6(8)9/h4-5H,3,7H2,1-2H3,(H,8,9)/t4-,5-/m0/s1

InChI key

AGPKZVBTJJNPAG-WHFBIAKZSA-N

General description

An essential branched chain amino acid.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
W527602I740356241
description

An essential branched chain amino acid.

description

99%, FCC, FG

description

from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%

description

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

form

crystals, crystalline solid

form

crystalline powder

form

powder

form

-

assay

≥99% (TLC)

assay

99%

assay

98.5-101.0%

assay

-

storage temp.

15-25°C

storage temp.

-

storage temp.

-

storage temp.

-

solubility

water: ~4% (at RT)

solubility

-

solubility

1 M HCl: 50 mg/mL

solubility

-

Quality Level

100

Quality Level

300

Quality Level

300

Quality Level

300

color

white

color

-

color

white

color

white


Still not finding the right product?

Explore all of our products under L-Isoleucine


Storage Class

13 - Non Combustible Solids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters