Skip to Content
Merck
All Photos(1)

Key Documents

410957

Sigma-Aldrich

3-Isobutyl-1-methylxanthine

≥98% (HPLC), solid, cAMP and cGMP phosphodiesterase inhibitor, Calbiochem®

Synonym(s):

3-Isobutyl-1-methylxanthine, IBMX

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N4O2
CAS Number:
Molecular Weight:
222.24
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

3-Isobutyl-1-methylxanthine, A cell-permeable, non-specific inhibitor of cAMP and cGMP phosphodiesterases (IC₅₀ = 2-50 µM).

Quality Level

description

RTECS - ZD8500000

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

ethanol: 10 mg/mL
DMSO: 100 mM
methanol: 50 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

[n]1(c2nc[nH]c2[c]([n]([c]1=O)C)=O)CC(C)C

InChI

1S/C10H14N4O2/c1-6(2)4-14-8-7(11-5-12-8)9(15)13(3)10(14)16/h5-6H,4H2,1-3H3,(H,11,12)

InChI key

APIXJSLKIYYUKG-UHFFFAOYSA-N

General description

A cell-permeable, non-specific inhibitor of cAMP and cGMP phosphodiesterases (IC50 = 2-50 µM). Also acts as an adenosine receptor antagonist. Reported to inhibit TNF-α expression in adipocyte precursor cells.
A non-specific, cell-permeable inhibitor of cAMP and cGMP phosphodiesterases (IC50 = 2-50 µM). Also acts as an adenosine receptor antagonist. Reported to inhibit TNFα expression in adipocyte precursor cells.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
cAMP and cGMP phosphodiesterases
Product does not compete with ATP.
Reversible: no
Target IC50: 2-50 µM against cAMP and cGMP phosphodiesterases

Warning

Toxicity: Standard Handling (A)

Preparation Note

Heating may be required to achieve complete solubilization.

Reconstitution

Following reconstitution in ethanol or methanol, refrigerate (4°C). Following reconstitution in DMSO, aliquot and freeze (-20°C). Ethanol/methanol and DMSO stock solutions are stable for up to 6 months at 4°C or -20°C, respectively.

Other Notes

Hube, F., et al. 1999. Horm. Metab. Res.31, 359.
Morgan, A.J., et al. 1993. Biochem. Pharmacol.45, 2373.
Scamps, F., et al. 1993. Eur. J. Pharmacol. 244, 119.
Turner, N.C., et al. 1993. Br. J. Pharmacol. 108, 876.
Tamura, T., et al. 1991. J. Gen. Physiol. 98, 95.
Beavo, J.A., and Reifsnyder, D.H. 1990. Trends Pharmacol. Sci.11, 150.
Russell, T.R. 1979. Proc. Natl. Acad. Sci. USA76, 4451.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiankun Xu et al.
Bioactive materials, 8, 95-108 (2021-09-21)
Magnesium metal and its alloys are being developed as effective orthopedic implants; however, the mechanisms underlying the actions of magnesium on bones remain unclear. Cystic fibrosis, the most common genetic disease in Caucasians caused by the mutation of CFTR, has
Jiang Li et al.
Antioxidants (Basel, Switzerland), 13(5) (2024-05-25)
Hypertension reduces the bioavailability of vascular nitric oxide (NO) and contributes to the onset of vascular dementia (VaD). A loss of NO bioavailability increases inflammation and oxidative stress. 2-(4-Methylthiazol-5-yl) ethyl nitrate hydrochloride (W1302) is a novel nitric oxide donor (NOD)
Salaheldeen Elsaid et al.
FEBS open bio, 14(8), 1277-1290 (2024-06-26)
In multicellular organisms, stem cells are impacted by microenvironmental resources such as nutrient availability and oxygen tension for their survival, growth, and differentiation. However, the accessibility of these resources in the pericellular environment greatly varies from organ to organ. This
Dina Hany et al.
Science advances, 9(19), eadd3685-eadd3685 (2023-05-12)
In breast cancer, resistance to endocrine therapies that target estrogen receptor α (ERα), such as tamoxifen and fulvestrant, remains a major clinical problem. Whether and how ERα+ breast cancers switch from being estrogen-dependent to estrogen-independent remains unclear. With a genome-wide
H Ji
Methods in molecular medicine, 51, 363-377 (2001-01-01)
Angiotensin type-1 receptors (AT(1) receptors) mediate various physiological actions of angiotensin (Ang II) via multiple-signal transduction pathways (1). In addition to the phospholipase C pathway and dihydropyridine-sensitive voltage-dependent calcium channels, AT(1) receptors can couple to inhibition of adenylate cyclase via

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service