Skip to Content
Merck

40-285

MILLIPLEX® Hand-Held Magnetic Plate Washer

This handheld magnetic plate washer efficiently removes liquid from 96-well plates for MILLIPLEX® assays by decanting or blotting.

Synonym(s):

Handheld plate washer, Magnetic bead washer, Manual assay washer, Multiplex bead wash tool

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41104923
NACRES:
NA.51
eCl@ss:
32039006
Manufacturer/tradename:
Milliplex®
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


manufacturer/tradename

Milliplex®

technique(s)

multiplexing: suitable

shipped in

ambient

General description

Extended Specifications
  • Plate Compatibility: 96‑well flat bottom plates
  • Description: Manual handheld washer for bead separation and washing
  • Application: Magnetic separation block for efficient bead capture and wash
  • Assay Compatibility: Fluorometric multiplexing assays
The Hand-Held Magnetic Plate Washer (Cat. No. 40-285) is a compact magnetic separator block designed for MILLIPLEX® assays. It enables efficient bead washing in 96‑well plates, ensuring reliable and reproducible assay performance.

Application

Designed for MILLIPLEX® multiplex magnetic bead-based assays.

Features and Benefits

  • Compact Design: The handheld magnetic separator block simplifies bead washing while ensuring reliable assay performance.
  • Optimized for MILLIPLEX® Assays: Specifically designed for multiplex assays, delivering consistent and reproducible results.
  • Versatile Compatibility: Compatible with 96-well flat bottom plates, offering flexibility across assay formats.
  • Cost-Effective Manual Operation: Provides a budget-friendly solution compared to automated washers, with easy setup and use.
  • Efficient Magetic Separation: Utilizes magnetic separation technology for effective bead capture and buffer exchange, preserving assay integrity.

Legal Information

MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

For Research Use Only. Not for use in diagnostic procedures




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library