401442
Goat Anti-Human IgG, γ-Chain Specific Alkaline Phosphatase Conjugate
liquid, Calbiochem®
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
NACRES:
NA.46
Recommended Products
biological source
goat
Quality Level
antibody form
affinity isolated antibody
antibody product type
secondary antibodies
clone
polyclonal
form
liquid
contains
≤0.1% sodium azide as preservative
manufacturer/tradename
Calbiochem®
storage condition
do not freeze
isotype
IgG
shipped in
wet ice
storage temp.
2-8°C
General description
Immunoaffinity purified goat polyclonal antibody conjugated to alkaline phosphatase. Recognizes human IgG γ-chain.
This Goat Anti-Human IgG, γ-Chain Specific Alk-Phos Conjugate is validated for use in Enzyme Immunoassay, Immunoelectrophoresis for the detection of Human IgG, γ-Chain Specific.
Immunogen
Human
Application
Enzyme Immunoassay (1:5000)
Immunoelectrophoresis (see comments)
Immunoelectrophoresis (see comments)
Packaging
Please refer to vial label for lot-specific concentration.
Warning
Toxicity: Standard Handling (A)
Physical form
In 50 mM Tris-HCl, 1 mM MgCl₂, 0.1 mM ZnCl₂, 1% BSA, pH 8.0.
Other Notes
Monospecific for human IgG γ-chain as determined by immunoelectrophoresis against normal human serum. p-Nitrophenylphosphate, 1 mg/ml in 10% diethanolamine, pH 9.8, 1 mM MgCl2 at 25°C, was used as substrate for testing immunoenzyme reactivity. Variables associated with assay conditions will dictate the proper working dilution.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Darren Suryawijaya Ong et al.
Nature communications, 15(1), 6968-6968 (2024-08-14)
The use of pneumococcal conjugate vaccine (PCV) schedules with fewer doses are being considered to reduce costs and improve access, particularly in low- and middle-income countries. While several studies have assessed their immunogenicity, there are limited data on their potential
Aapo Knuutila et al.
Emerging microbes & infections, 12(1), 2204146-2204146 (2023-04-16)
Immunization during pregnancy (IP) against pertussis is recommended in many countries to protect infants. Although maternal antibodies can influence the infants' antibody responses to primary vaccinations, their effect on the development of functional antibodies and B cells remain poorly studied.
Pauline Versteegen et al.
Frontiers in immunology, 13, 864674-864674 (2022-06-10)
Immunogenicity of acellular pertussis (aP) vaccines is conventionally assessed by measuring antibody responses but antibody concentrations wane quickly after vaccination. Memory B cells, however, are critical in sustaining long-term protection and therefore may be an important factor when assessing pertussis
Jordan R Barrett et al.
Frontiers in immunology, 14, 1193079-1193079 (2024-02-01)
We have previously reported primary endpoints of a clinical trial testing two vaccine platforms for the delivery of Plasmodium vivax malaria DBPRII: viral vectors (ChAd63, MVA), and protein/adjuvant (PvDBPII with 50µg Matrix-M™ adjuvant). Delayed boosting was necessitated due to trial
Malick M Gibani et al.
Nature medicine, 25(7), 1082-1088 (2019-07-05)
Salmonella Typhi is a human host-restricted pathogen that is responsible for typhoid fever in approximately 10.9 million people annually1. The typhoid toxin is postulated to have a central role in disease pathogenesis, the establishment of chronic infection and human host
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service