Skip to Content
Merck
All Photos(1)

Key Documents

401215

Sigma-Aldrich

Goat Anti-Mouse IgG, H&L Chain Specific Peroxidase Conjugate

liquid, Calbiochem®

Synonym(s):

Goat IgG antibody, Goat anti-mouse IgG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

Quality Level

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

liquid

contains

≤0.02% Thimerosal as preservative

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Related Categories

General description

Immunoaffinity purified goat polyclonal antibody conjugated to horseradish peroxidase (HRP). Recognizes mouse IgG heavy and light chains.
This Goat Anti-Mouse IgG, H&L Chain Specific Peroxidase Conjugate is validated for use in Enzyme Immunoassay, Immunoelectrophoresis for the detection of Mouse IgG, H&L Chain Specific.

Application

Enzyme Immunoassay (1:10,000)

Immunoelectrophoresis (see comments)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In PBS, 0.5 mM EDTA, 10% mannose, 2% sucrose, 1% BSA, pH 7.6.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

To comply with ban of sale of mercury-added products required by The Interstate Mercury Education and Reduction Clearinghouse (IMERC), this product is prohibited to be sold in the following US states: Rhode Island and Connecticut.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyo-Seok Seo et al.
Biomolecules & therapeutics, 22(6), 525-531 (2014-12-10)
In the present study, we investigated whether apigenin significantly affects tumor necrosis factor-α (TNF-α)-induced production and gene expression of MUC5AC mucin in airway epithelial cells. Confluent NCI-H292 cells were pretreated with apigenin for 30 min and then stimulated with TNF-α
Yuji Goto et al.
Oncotarget, 7(3), 2379-2390 (2015-10-28)
Late SV40 factor 3 (LSF), a transcription factor, contributes to human hepatocellular carcinoma (HCC). However, decreased expression level of LSF in skin melanoma compared to that in benign melanocytic tumors and nevi in mice and humans was found in this
András Zeke et al.
Molecular systems biology, 11(11), 837-837 (2015-11-06)
Mitogen-activated protein kinases (MAPK) are broadly used regulators of cellular signaling. However, how these enzymes can be involved in such a broad spectrum of physiological functions is not understood. Systematic discovery of MAPK networks both experimentally and in silico has
Serena Notartomaso et al.
Scientific reports, 8(1), 13361-13361 (2018-09-08)
In cerebellar Purkinje cells (PCs) type-1 metabotropic glutamate (mGlu1) receptors play a key role in motor learning and drive the refinement of synaptic innervation during postnatal development. The cognate mGlu5 receptor is absent in mature PCs and shows low expression
Chunsheng Liu et al.
Circulation research, 97(2), 135-143 (2005-06-25)
Recent experimental studies suggest C-reactive protein (CRP) may be a potential mediator of atherosclerosis and its complications. However, there is growing criticism of in vitro CRP studies that use commercial CRP preparations containing biologically active contaminants. The effects of commercial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service