Skip to Content
Merck

357002

Glycine, Molecular Biology Grade

Absorbance (260 nm and 280 nm): ≤0.01.

Synonym(s):

Glycine, Aminoacetic acid, Aminoethanoic acid, Glycocoll

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
NH2CH2COOH
CAS Number:
Molecular Weight:
75.07
NACRES:
NA.28
UNSPSC Code:
12352209
Beilstein/REAXYS Number:
635782
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

Molecular Biology

Quality Segment

description

Merck USA index - 14, 4491

assay

≥99% (titration)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

pKa (25 °C)

(1) 2.35, (2) 9.60, 2.35

mp

240 °C (dec.) (lit.)

solubility

water: 200 mg/mL

foreign activity

DNases, proteases, RNases, none detected

shipped in

ambient

storage temp.

15-25°C

SMILES string

NCC(O)=O

InChI

1S/C2H5NO2/c3-1-2(4)5/h1,3H2,(H,4,5)

InChI key

DHMQDGOQFOQNFH-UHFFFAOYSA-N

General description

A non-essential amino acid that can be used in the preparation of biological buffers (absorbance: ≤0.01 at 260 nm and 280 nm).

Biochem/physiol Actions

Inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.

Preparation Note

Following reconstitution, store at room temperature (20°C). Stock solutions are stable for up to 6 months at room temperature.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

13 - Non Combustible Solids

wgk

WGK 1

flash_point_f

No data available

flash_point_c

No data available



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library