Skip to Content
Merck
All Photos(1)

Key Documents

341600

Sigma-Aldrich

Fibrinogen, Fragment D, Human Plasma

Fibrinogen fragment D is a thermolabile native human plasma protein prepared by digestion of purified fibrinogen followed by preparative electrophoresis.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.41

Quality Level

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤1% Fragment E

solubility

sterile distilled water: 200 μg/mL

shipped in

ambient

storage temp.

−20°C

General description

Native fibrinogen fragment D from human plasma prepared by plasmin digestion of purified fibrinogen followed by preparative electrophoresis. Thermolabile fragment. Complete proteolysis of a single fibrinogen molecule produces two D and one E fragments. Fragment D has been shown to increase endothelial monolayer permeability and to decrease polymorphonuclear leukocyte adhesion to endothelial cells. The D and E fragments have no common antigenic determinants; therefore, no cross-reactivity occurs.

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 400 mM glycine, 150 mM NaCl.

Reconstitution

Following reconstitution, aliquot and freeze (-70°C). Stock solutions are stable for up to 4 h at 20°C or for up to 8 h at 4°C or for up to 3 months at -70°C.

Analysis Note

single band by SDS-PAGE

Other Notes

Spraggon, G., et al. 1997. Nature 389, 455.
Stirk, C.M., et al. 1993. Atherosclerosis103, 159.
Vogel, P., et al. 1993. Eur. Surg. Res.25, 83.
Azpiazu, I., and Chapman, D. 1992. Biochim. Biophys. Acta1119, 268.
Thompson, W.D., et al. 1992. J. Pathol.168, 47.
Fisher, E.G., et al. 1991. Haemostasis21, 141.
Ge, M., et al. 1991. Am. J. Physiol.261, L283.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service