Skip to Content
Merck
All Photos(1)

Key Documents

251600

Sigma-Aldrich

Daidzein

Inactive analog of Genistein.

Synonym(s):

Daidzein, 4ʹ,7-Dihydroxyisoflavone, Casein Kinase II Inhibitor X

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10O4
CAS Number:
Molecular Weight:
254.24
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

description

Merck USA index - 14, 2801

Assay

≥98% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

off-white

solubility

DMSO: 200 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

O1c2c(ccc(c2)O)C(=O)C(=C1)c3ccc(cc3)O

InChI

1S/C15H10O4/c16-10-3-1-9(2-4-10)13-8-19-14-7-11(17)5-6-12(14)15(13)18/h1-8,16-17H

InChI key

ZQSIJRDFPHDXIC-UHFFFAOYSA-N

General description

Inactive analog of Genistein (Cat. No. 345834). Blocks the G1 phase of the cell cycle in Swiss/3T3 cells by inhibiting casein kinase II activity. Inhibits the growth of MiaPaC-2 and PANC-1 cells (IC50= 45 µM and 75 µM, respectively).
Inactive analog of genistein (Cat. No. 345834). Blocks the G1 phase of the cell cycle in Swiss 3T3 cells by inhibiting casein kinase II activity. Inhibits epidermal growth factor (EGF) receptor autophosphorylation (IC50 >300 µM). Inhibits the growth of MiaPaC-2 and PANC-1 cells (IC50= 45 µM and 75 µM, respectively).

Biochem/physiol Actions

Cell permeable: no
Primary Target
MiaPaC-2, PANC-1
Product does not compete with ATP.
Reversible: no
Target IC50: 45 µM, 75 µM for inhibiting the growth of MiaPaC-2 and PANC-1 cells, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C) under an inert gas. DMSO stock solutions are stable for up to 2 months at -20°C.

Other Notes

Guo, J-M., et al. 2004. World J. Gastroenterol.10, 860.
Higashi, K., and Ogawara, H. 1994. Biochim. Biophys. Acta 1221, 29.
Lavens, S.E., et al. 1992. Am. J. Respir. Cell Mol. Biol.7, 637.
Akiyama, T., et al. 1987. J. Biol. Chem.262, 5592.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service